Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 77297875-77298100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK5
Entrez Gene ID
26289
Gene Name
adenylate kinase 5
Gene Aliases
AK6
UniGene
Hs.559718
Gene Chromosome Location
Chr.1: 77281908-77559969 on Build GRCh38
Transcripts
NM_012093.3 NM_174858.2 XM_005270739.4 XM_006710572.3 XM_017001008.1 XM_017001009.1 XM_017001010.1 XM_017001011.1 XM_017001012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6597348 c.655G>T p.G219* Chr.1: 77297903-77297903 on Build GRCh38
2242292 c.674C>G p.A225G Chr.1: 77297922-77297922 on Build GRCh38
4660705 c.630T>C p.P210P Chr.1: 77297878-77297878 on Build GRCh38
6024755 c.695A>G p.D232G Chr.1: 77297943-77297943 on Build GRCh38
2242293 c.686C>A p.S229Y Chr.1: 77297934-77297934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113646551 Chr.1: 77297948-77297948 on Build GRCh38
rs1020219103 Chr.1: 77298058-77298058 on Build GRCh38
rs767519551 Chr.1: 77297987-77297987 on Build GRCh38
rs768069035 Chr.1: 77297981-77297981 on Build GRCh38
rs899948220 Chr.1: 77297941-77297941 on Build GRCh38
rs759686268 Chr.1: 77297959-77297959 on Build GRCh38
rs747578077 Chr.1: 77298075-77298075 on Build GRCh38
rs866054610 Chr.1: 77297913-77297913 on Build GRCh38
rs956737871 Chr.1: 77298002-77298002 on Build GRCh38
rs1040964894 Chr.1: 77298023-77298023 on Build GRCh38
rs769976107 Chr.1: 77297964-77297964 on Build GRCh38
rs747195859 Chr.1: 77297981-77297987 on Build GRCh38
rs773867163 Chr.1: 77298035-77298035 on Build GRCh38
rs772617454 Chr.1: 77297902-77297902 on Build GRCh38
rs775757390 Chr.1: 77297974-77297974 on Build GRCh38
rs773420804 Chr.1: 77297917-77297917 on Build GRCh38
rs771224866 Chr.1: 77297947-77297947 on Build GRCh38
rs866129991 Chr.1: 77298080-77298080 on Build GRCh38
rs978593474 Chr.1: 77298076-77298076 on Build GRCh38
rs1029700613 Chr.1: 77297993-77297993 on Build GRCh38
rs747361184 Chr.1: 77297942-77297942 on Build GRCh38
rs1010527508 Chr.1: 77298056-77298056 on Build GRCh38
rs748642477 Chr.1: 77297901-77297901 on Build GRCh38
rs1039783303 Chr.1: 77297918-77297918 on Build GRCh38

View Full Product Details