Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62604686-62604915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL3
Entrez Gene ID
27329
Gene Name
angiopoietin like 3
Gene Aliases
ANG-5, ANGPT5, ANL3, FHBL2
UniGene
Hs.209153
Gene Chromosome Location
Chr.1: 62597487-62606305 on Build GRCh38
Transcripts
NM_014495.3 NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_001272002.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_011542330.2 XM_017002639.1 XM_017002640.1

Gene Symbol
DOCK7
Entrez Gene ID
85440
Gene Name
dedicator of cytokinesis 7
Gene Aliases
EIEE23, ZIR2
UniGene
Hs.744927
Gene Chromosome Location
Chr.1: 62454726-62688368 on Build GRCh38
Transcripts
NM_014495.3 NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_001272002.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_011542330.2 XM_017002639.1 XM_017002640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4009259 c.1297T>C p.S433P Chr.1: 62604731-62604731 on Build GRCh38
6527631 c.1363G>C p.D455H Chr.1: 62604797-62604797 on Build GRCh38
6851717 c.1289G>A p.R430K Chr.1: 62604723-62604723 on Build GRCh38
6560903 c.1350G>C p.L450F Chr.1: 62604784-62604784 on Build GRCh38
7237082 c.1279G>C p.E427Q Chr.1: 62604713-62604713 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781279447 Chr.1: 62604774-62604774 on Build GRCh38
rs753230849 Chr.1: 62604749-62604749 on Build GRCh38
rs375746080 Chr.1: 62604859-62604859 on Build GRCh38
rs759010000 Chr.1: 62604754-62604754 on Build GRCh38
rs67461605 Chr.1: 62604867-62604875 on Build GRCh38
rs775677524 Chr.1: 62604866-62604867 on Build GRCh38
rs753454044 Chr.1: 62604838-62604838 on Build GRCh38
rs760361956 Chr.1: 62604716-62604716 on Build GRCh38
rs765960809 Chr.1: 62604719-62604719 on Build GRCh38
rs776851296 Chr.1: 62604868-62604869 on Build GRCh38
rs753175818 Chr.1: 62604864-62604864 on Build GRCh38
rs745852131 Chr.1: 62604764-62604769 on Build GRCh38
rs759309147 Chr.1: 62604857-62604857 on Build GRCh38
rs940005040 Chr.1: 62604760-62604760 on Build GRCh38
rs760107372 Chr.1: 62604819-62604819 on Build GRCh38
rs143404279 Chr.1: 62604799-62604799 on Build GRCh38
rs749942909 Chr.1: 62604713-62604713 on Build GRCh38
rs771131159 Chr.1: 62604770-62604770 on Build GRCh38
rs34483103 Chr.1: 62604869-62604877 on Build GRCh38
rs201148287 Chr.1: 62604820-62604820 on Build GRCh38
rs754163855 Chr.1: 62604720-62604720 on Build GRCh38
rs777991106 Chr.1: 62604761-62604761 on Build GRCh38
rs772720883 Chr.1: 62604815-62604815 on Build GRCh38
rs766013795 Chr.1: 62604821-62604821 on Build GRCh38
rs760709309 Chr.1: 62604866-62604874 on Build GRCh38
rs771856463 Chr.1: 62604769-62604770 on Build GRCh38
rs775478158 Chr.1: 62604794-62604795 on Build GRCh38
rs761708356 Chr.1: 62604813-62604813 on Build GRCh38
rs748938254 Chr.1: 62604715-62604717 on Build GRCh38
rs755423135 Chr.1: 62604732-62604732 on Build GRCh38
rs749073340 Chr.1: 62604791-62604791 on Build GRCh38
rs774181918 Chr.1: 62604801-62604801 on Build GRCh38
rs778508040 Chr.1: 62604764-62604767 on Build GRCh38
rs758956598 Chr.1: 62604867-62604867 on Build GRCh38
rs770867880 Chr.1: 62604762-62604763 on Build GRCh38
rs908659981 Chr.1: 62604748-62604748 on Build GRCh38

View Full Product Details