Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151745456-151745727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL2
Entrez Gene ID
344752
Gene Name
arylacetamide deacetylase like 2
Gene Aliases
-
UniGene
Hs.144710
Gene Chromosome Location
Chr.3: 151733916-151758016 on Build GRCh38
Transcripts
NM_207365.3 XM_011512753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1717907 c.506G>A p.G169E Chr.3: 151745649-151745649 on Build GRCh38
7086554 c.441delG p.E148fs*25 Chr.3: 151745584-151745584 on Build GRCh38
5398422 c.412G>A p.G138S Chr.3: 151745555-151745555 on Build GRCh38
7086624 c.443A>T p.E148V Chr.3: 151745586-151745586 on Build GRCh38
3748805 c.373C>T p.L125L Chr.3: 151745516-151745516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761503526 Chr.3: 151745617-151745617 on Build GRCh38
rs149013573 Chr.3: 151745536-151745536 on Build GRCh38
rs764915485 Chr.3: 151745700-151745700 on Build GRCh38
rs777744746 Chr.3: 151745643-151745643 on Build GRCh38
rs142920843 Chr.3: 151745541-151745541 on Build GRCh38
rs756826800 Chr.3: 151745519-151745519 on Build GRCh38
rs749355039 Chr.3: 151745644-151745644 on Build GRCh38
rs774615903 Chr.3: 151745608-151745608 on Build GRCh38
rs766866019 Chr.3: 151745592-151745592 on Build GRCh38
rs751412229 Chr.3: 151745629-151745629 on Build GRCh38
rs151100169 Chr.3: 151745579-151745579 on Build GRCh38
rs748293123 Chr.3: 151745564-151745564 on Build GRCh38
rs201615784 Chr.3: 151745565-151745565 on Build GRCh38
rs779746188 Chr.3: 151745554-151745554 on Build GRCh38
rs200688205 Chr.3: 151745615-151745615 on Build GRCh38
rs778424556 Chr.3: 151745522-151745522 on Build GRCh38
rs370809556 Chr.3: 151745648-151745648 on Build GRCh38
rs199837914 Chr.3: 151745494-151745494 on Build GRCh38
rs761593326 Chr.3: 151745684-151745684 on Build GRCh38
rs199625823 Chr.3: 151745547-151745547 on Build GRCh38
rs908140625 Chr.3: 151745594-151745594 on Build GRCh38
rs773281027 Chr.3: 151745572-151745572 on Build GRCh38
rs763908870 Chr.3: 151745614-151745614 on Build GRCh38
rs35364301 Chr.3: 151745624-151745625 on Build GRCh38
rs760055575 Chr.3: 151745667-151745667 on Build GRCh38
rs764675842 Chr.3: 151745619-151745619 on Build GRCh38
rs370659768 Chr.3: 151745701-151745701 on Build GRCh38
rs945745607 Chr.3: 151745578-151745578 on Build GRCh38
rs138796090 Chr.3: 151745634-151745634 on Build GRCh38
rs987137633 Chr.3: 151745567-151745567 on Build GRCh38
rs772048754 Chr.3: 151745694-151745695 on Build GRCh38
rs1972977 Chr.3: 151745633-151745633 on Build GRCh38
rs745342379 Chr.3: 151745491-151745491 on Build GRCh38
rs780021458 Chr.3: 151745622-151745622 on Build GRCh38
rs746776640 Chr.3: 151745556-151745556 on Build GRCh38
rs763806160 Chr.3: 151745487-151745487 on Build GRCh38
rs772579314 Chr.3: 151745668-151745668 on Build GRCh38
rs199875931 Chr.3: 151745635-151745635 on Build GRCh38
rs759968096 Chr.3: 151745611-151745611 on Build GRCh38
rs370207359 Chr.3: 151745641-151745641 on Build GRCh38
rs770009039 Chr.3: 151745636-151745636 on Build GRCh38
rs112885334 Chr.3: 151745502-151745502 on Build GRCh38
rs912867101 Chr.3: 151745574-151745574 on Build GRCh38
rs375247147 Chr.3: 151745652-151745652 on Build GRCh38
rs768913808 Chr.3: 151745559-151745559 on Build GRCh38
rs776818363 Chr.3: 151745560-151745560 on Build GRCh38
rs1550964 Chr.3: 151745705-151745705 on Build GRCh38
rs141274337 Chr.3: 151745621-151745621 on Build GRCh38

View Full Product Details