Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7852919-7853192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI5L
Entrez Gene ID
115704
Gene Name
ecotropic viral integration site 5 like
Gene Aliases
-
UniGene
Hs.26870
Gene Chromosome Location
Chr.19: 7830275-7864976 on Build GRCh38
Transcripts
NM_001159944.2 NM_145245.4 XM_005272458.4 XM_017026239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1305309 c.1014G>C p.Q338H Chr.19: 7853112-7853112 on Build GRCh38
6656921 c.1015T>A p.F339I Chr.19: 7853113-7853113 on Build GRCh38
3990410 c.1006C>G p.P336A Chr.19: 7853104-7853104 on Build GRCh38
6656918 c.1017C>T p.F339F Chr.19: 7853115-7853115 on Build GRCh38
6344135 c.1085+7G>T p.? Chr.19: 7853190-7853190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376219402 Chr.19: 7853066-7853066 on Build GRCh38
rs376830712 Chr.19: 7853037-7853037 on Build GRCh38
rs372805600 Chr.19: 7853034-7853034 on Build GRCh38
rs529831805 Chr.19: 7853004-7853004 on Build GRCh38
rs781446338 Chr.19: 7853142-7853142 on Build GRCh38
rs959462084 Chr.19: 7852974-7852974 on Build GRCh38
rs990890817 Chr.19: 7852982-7852982 on Build GRCh38
rs773502662 Chr.19: 7853070-7853070 on Build GRCh38
rs763482170 Chr.19: 7853072-7853072 on Build GRCh38
rs375217848 Chr.19: 7853127-7853127 on Build GRCh38
rs946929167 Chr.19: 7853008-7853008 on Build GRCh38
rs141315535 Chr.19: 7853165-7853165 on Build GRCh38
rs150896280 Chr.19: 7853118-7853118 on Build GRCh38
rs1048939740 Chr.19: 7852998-7852999 on Build GRCh38
rs112306213 Chr.19: 7853023-7853023 on Build GRCh38
rs748489666 Chr.19: 7853074-7853074 on Build GRCh38
rs929129144 Chr.19: 7852992-7852992 on Build GRCh38
rs769953060 Chr.19: 7853151-7853151 on Build GRCh38
rs192711713 Chr.19: 7852947-7852947 on Build GRCh38
rs763926544 Chr.19: 7853115-7853115 on Build GRCh38
rs768872477 Chr.19: 7853048-7853048 on Build GRCh38
rs1032544792 Chr.19: 7853059-7853059 on Build GRCh38
rs546678983 Chr.19: 7853005-7853005 on Build GRCh38
rs1002167481 Chr.19: 7853014-7853014 on Build GRCh38
rs368326668 Chr.19: 7853069-7853069 on Build GRCh38
rs760065192 Chr.19: 7853172-7853172 on Build GRCh38
rs866587091 Chr.19: 7852956-7852956 on Build GRCh38
rs12975442 Chr.19: 7853033-7853033 on Build GRCh38
rs61744445 Chr.19: 7853109-7853109 on Build GRCh38
rs767018098 Chr.19: 7853103-7853103 on Build GRCh38
rs919061216 Chr.19: 7852991-7852991 on Build GRCh38
rs745805699 Chr.19: 7853132-7853132 on Build GRCh38
rs753966562 Chr.19: 7853064-7853064 on Build GRCh38
rs773759413 Chr.19: 7852963-7852965 on Build GRCh38
rs1040174495 Chr.19: 7852979-7852979 on Build GRCh38
rs1013595736 Chr.19: 7853029-7853029 on Build GRCh38
rs748363489 Chr.19: 7853144-7853144 on Build GRCh38
rs558062912 Chr.19: 7853050-7853050 on Build GRCh38
rs571945687 Chr.19: 7853060-7853060 on Build GRCh38
rs113867846 Chr.19: 7853077-7853077 on Build GRCh38
rs371830480 Chr.19: 7853079-7853079 on Build GRCh38
rs200315839 Chr.19: 7853152-7853152 on Build GRCh38
rs887684825 Chr.19: 7853001-7853001 on Build GRCh38
rs919020490 Chr.19: 7853137-7853137 on Build GRCh38
rs896520690 Chr.19: 7853027-7853027 on Build GRCh38
rs755071695 Chr.19: 7853140-7853140 on Build GRCh38
rs766858322 Chr.19: 7853076-7853076 on Build GRCh38
rs755263820 Chr.19: 7853003-7853003 on Build GRCh38
rs774770123 Chr.19: 7853169-7853169 on Build GRCh38
rs771337787 Chr.19: 7853166-7853166 on Build GRCh38
rs957026877 Chr.19: 7852954-7852954 on Build GRCh38
rs753644059 Chr.19: 7853116-7853116 on Build GRCh38

View Full Product Details