Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 26508211-26508480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
UniGene
Hs.577269
Gene Chromosome Location
Chr.11: 26189123-26663289 on Build GRCh38
Transcripts
NM_001313726.1 NM_001313727.1 NM_031418.3 XM_017018118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5676413 c.544A>G p.T182A Chr.11: 26508215-26508215 on Build GRCh38
4032160 c.591G>A p.E197E Chr.11: 26508262-26508262 on Build GRCh38
2113041 c.559C>T p.L187F Chr.11: 26508230-26508230 on Build GRCh38
6873213 c.591+1G>A p.? Chr.11: 26508263-26508263 on Build GRCh38
7421057 c.582G>C p.L194F Chr.11: 26508253-26508253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577138251 Chr.11: 26508326-26508326 on Build GRCh38
rs117136103 Chr.11: 26508336-26508336 on Build GRCh38
rs756436710 Chr.11: 26508317-26508317 on Build GRCh38
rs149415582 Chr.11: 26508237-26508237 on Build GRCh38
rs1018726020 Chr.11: 26508442-26508442 on Build GRCh38
rs796821880 Chr.11: 26508372-26508372 on Build GRCh38
rs951674802 Chr.11: 26508270-26508270 on Build GRCh38
rs768733707 Chr.11: 26508334-26508334 on Build GRCh38
rs751046428 Chr.11: 26508297-26508297 on Build GRCh38
rs926752002 Chr.11: 26508299-26508299 on Build GRCh38
rs771260539 Chr.11: 26508325-26508325 on Build GRCh38
rs191832866 Chr.11: 26508423-26508423 on Build GRCh38
rs757979465 Chr.11: 26508275-26508275 on Build GRCh38
rs777996331 Chr.11: 26508319-26508319 on Build GRCh38
rs294025 Chr.11: 26508353-26508353 on Build GRCh38
rs750021659 Chr.11: 26508273-26508273 on Build GRCh38
rs200157589 Chr.11: 26508243-26508243 on Build GRCh38
rs982687678 Chr.11: 26508348-26508348 on Build GRCh38
rs371337106 Chr.11: 26508312-26508312 on Build GRCh38
rs749586011 Chr.11: 26508321-26508321 on Build GRCh38
rs1052648401 Chr.11: 26508368-26508368 on Build GRCh38
rs889180087 Chr.11: 26508427-26508427 on Build GRCh38
rs893830000 Chr.11: 26508400-26508400 on Build GRCh38
rs755655912 Chr.11: 26508340-26508340 on Build GRCh38
rs763746681 Chr.11: 26508349-26508349 on Build GRCh38
rs1041081282 Chr.11: 26508366-26508366 on Build GRCh38
rs755517489 Chr.11: 26508311-26508311 on Build GRCh38
rs375570481 Chr.11: 26508266-26508266 on Build GRCh38
rs745905852 Chr.11: 26508329-26508329 on Build GRCh38
rs118043383 Chr.11: 26508441-26508441 on Build GRCh38
rs564399403 Chr.11: 26508412-26508412 on Build GRCh38
rs118181050 Chr.11: 26508314-26508314 on Build GRCh38
rs541564673 Chr.11: 26508240-26508240 on Build GRCh38
rs541340564 Chr.11: 26508281-26508281 on Build GRCh38

View Full Product Details