Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93993104-93993375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA4
Entrez Gene ID
24
Gene Name
ATP binding cassette subfamily A member 4
Gene Aliases
ABC10, ABCR, ARMD2, CORD3, FFM, RMP, RP19, STGD, STGD1
UniGene
Hs.416707
Gene Chromosome Location
Chr.1: 93992837-94121149 on Build GRCh38
Transcripts
NM_000350.2

SNP Detail

SNP ID Chromosome Location
rs35896233 Chr.1: 93993126-93993127 on Build GRCh38
rs767072316 Chr.1: 93993193-93993193 on Build GRCh38
rs370860160 Chr.1: 93993267-93993267 on Build GRCh38
rs377008865 Chr.1: 93993226-93993226 on Build GRCh38
rs749875918 Chr.1: 93993198-93993198 on Build GRCh38
rs755828662 Chr.1: 93993203-93993203 on Build GRCh38
rs201160433 Chr.1: 93993182-93993182 on Build GRCh38
rs577038211 Chr.1: 93993335-93993335 on Build GRCh38
rs75999881 Chr.1: 93993327-93993327 on Build GRCh38
rs375731558 Chr.1: 93993269-93993269 on Build GRCh38
rs955079945 Chr.1: 93993221-93993221 on Build GRCh38
rs866960838 Chr.1: 93993270-93993270 on Build GRCh38
rs902542806 Chr.1: 93993139-93993139 on Build GRCh38
rs775865861 Chr.1: 93993243-93993243 on Build GRCh38
rs765812753 Chr.1: 93993283-93993283 on Build GRCh38
rs753523367 Chr.1: 93993286-93993286 on Build GRCh38
rs145634012 Chr.1: 93993227-93993227 on Build GRCh38
rs200205293 Chr.1: 93993277-93993277 on Build GRCh38
rs762183450 Chr.1: 93993188-93993188 on Build GRCh38
rs112217438 Chr.1: 93993153-93993153 on Build GRCh38
rs767876749 Chr.1: 93993268-93993268 on Build GRCh38
rs748141123 Chr.1: 93993224-93993224 on Build GRCh38
rs200401067 Chr.1: 93993214-93993214 on Build GRCh38
rs945851538 Chr.1: 93993348-93993348 on Build GRCh38
rs769173189 Chr.1: 93993260-93993260 on Build GRCh38
rs754777067 Chr.1: 93993220-93993220 on Build GRCh38
rs575090193 Chr.1: 93993238-93993238 on Build GRCh38
rs766623453 Chr.1: 93993176-93993176 on Build GRCh38
rs1042423980 Chr.1: 93993132-93993132 on Build GRCh38
rs965884162 Chr.1: 93993130-93993130 on Build GRCh38
rs369306889 Chr.1: 93993222-93993222 on Build GRCh38
rs367853124 Chr.1: 93993275-93993275 on Build GRCh38
rs201835773 Chr.1: 93993185-93993185 on Build GRCh38
rs754576845 Chr.1: 93993289-93993289 on Build GRCh38
rs368493917 Chr.1: 93993303-93993303 on Build GRCh38
rs376708072 Chr.1: 93993261-93993261 on Build GRCh38
rs111435203 Chr.1: 93993133-93993133 on Build GRCh38
rs919146588 Chr.1: 93993162-93993162 on Build GRCh38
rs372398535 Chr.1: 93993225-93993225 on Build GRCh38
rs200946608 Chr.1: 93993211-93993211 on Build GRCh38
rs1035317143 Chr.1: 93993155-93993155 on Build GRCh38
rs962505043 Chr.1: 93993183-93993183 on Build GRCh38
rs946762540 Chr.1: 93993129-93993129 on Build GRCh38
rs534777895 Chr.1: 93993141-93993141 on Build GRCh38
rs749454214 Chr.1: 93993259-93993259 on Build GRCh38

View Full Product Details