Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41612764-41613037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAT4
Entrez Gene ID
137964
Gene Name
glycerol-3-phosphate acyltransferase 4
Gene Aliases
1-AGPAT 6, AGPAT6, LPAAT-zeta, LPAATZ, TSARG7
UniGene
Hs.355753
Gene Chromosome Location
Chr.8: 41578188-41625001 on Build GRCh38
Transcripts
NM_178819.3 XM_011544392.2 XM_011544393.1 XM_011544394.1 XM_017013042.1 XM_017013043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777403 c.832C>T p.Q278* Chr.8: 41612881-41612881 on Build GRCh38
287757 c.826G>T p.V276L Chr.8: 41612875-41612875 on Build GRCh38
354644 c.890A>T p.D297V Chr.8: 41612939-41612939 on Build GRCh38
2961262 c.823G>T p.G275C Chr.8: 41612872-41612872 on Build GRCh38
3649164 c.895C>T p.H299Y Chr.8: 41612944-41612944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039459573 Chr.8: 41612970-41612970 on Build GRCh38
rs765318607 Chr.8: 41612888-41612888 on Build GRCh38
rs771147289 Chr.8: 41612974-41612974 on Build GRCh38
rs765075072 Chr.8: 41612871-41612871 on Build GRCh38
rs115518769 Chr.8: 41612821-41612821 on Build GRCh38
rs779691963 Chr.8: 41612988-41612988 on Build GRCh38
rs552480796 Chr.8: 41612987-41612987 on Build GRCh38
rs761755761 Chr.8: 41612868-41612868 on Build GRCh38
rs754488446 Chr.8: 41612920-41612920 on Build GRCh38
rs372694683 Chr.8: 41612838-41612838 on Build GRCh38
rs770067704 Chr.8: 41612819-41612819 on Build GRCh38
rs977247047 Chr.8: 41612807-41612807 on Build GRCh38
rs778109201 Chr.8: 41612961-41612961 on Build GRCh38
rs774741159 Chr.8: 41612981-41612981 on Build GRCh38
rs777145697 Chr.8: 41612805-41612805 on Build GRCh38
rs980512939 Chr.8: 41612812-41612812 on Build GRCh38
rs763221766 Chr.8: 41612831-41612831 on Build GRCh38
rs201491109 Chr.8: 41612799-41612799 on Build GRCh38
rs758131155 Chr.8: 41612796-41612796 on Build GRCh38
rs780622204 Chr.8: 41612925-41612925 on Build GRCh38
rs374501472 Chr.8: 41612995-41612995 on Build GRCh38
rs1027412826 Chr.8: 41612792-41612792 on Build GRCh38
rs371903817 Chr.8: 41612922-41612922 on Build GRCh38
rs371678587 Chr.8: 41612798-41612798 on Build GRCh38
rs770484035 Chr.8: 41612801-41612801 on Build GRCh38
rs368929609 Chr.8: 41612900-41612900 on Build GRCh38
rs774689857 Chr.8: 41612839-41612839 on Build GRCh38
rs199736328 Chr.8: 41612928-41612928 on Build GRCh38
rs149518719 Chr.8: 41612984-41612984 on Build GRCh38
rs772296591 Chr.8: 41613010-41613010 on Build GRCh38
rs200257329 Chr.8: 41612907-41612907 on Build GRCh38
rs750301619 Chr.8: 41612893-41612893 on Build GRCh38
rs751164953 Chr.8: 41612911-41612911 on Build GRCh38
rs775773492 Chr.8: 41613011-41613011 on Build GRCh38
rs368508983 Chr.8: 41612797-41612797 on Build GRCh38
rs748587561 Chr.8: 41612816-41612816 on Build GRCh38
rs372558815 Chr.8: 41612853-41612853 on Build GRCh38
rs746385673 Chr.8: 41612795-41612796 on Build GRCh38
rs376290343 Chr.8: 41612859-41612859 on Build GRCh38
rs770397529 Chr.8: 41612941-41612941 on Build GRCh38

View Full Product Details