Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10574787-10575058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M2
Entrez Gene ID
10053
Gene Name
adaptor related protein complex 1 mu 2 subunit
Gene Aliases
AP1-mu2, HSMU1B, MU-1B, MU1B, mu2
UniGene
Hs.18894
Gene Chromosome Location
Chr.19: 10572671-10587315 on Build GRCh38
Transcripts
NM_001300887.1 NM_005498.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611500 c.1130G>T p.G377V Chr.19: 10574947-10574947 on Build GRCh38
5464380 c.1076G>A p.R359Q Chr.19: 10575007-10575007 on Build GRCh38
1611501 c.1129G>T p.G377W Chr.19: 10574948-10574948 on Build GRCh38
5758581 c.1159G>A p.V387I Chr.19: 10574918-10574918 on Build GRCh38
4662226 c.1126A>G p.I376V Chr.19: 10574951-10574951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769271733 Chr.19: 10574841-10574841 on Build GRCh38
rs762549075 Chr.19: 10574929-10574929 on Build GRCh38
rs752809774 Chr.19: 10574899-10574899 on Build GRCh38
rs956012083 Chr.19: 10574813-10574813 on Build GRCh38
rs767976584 Chr.19: 10574888-10574888 on Build GRCh38
rs987438798 Chr.19: 10574821-10574821 on Build GRCh38
rs924659026 Chr.19: 10574807-10574807 on Build GRCh38
rs745694140 Chr.19: 10574952-10574952 on Build GRCh38
rs2304162 Chr.19: 10574833-10574833 on Build GRCh38
rs757472950 Chr.19: 10575025-10575025 on Build GRCh38
rs768006330 Chr.19: 10574990-10574990 on Build GRCh38
rs1038872436 Chr.19: 10574854-10574854 on Build GRCh38
rs779076082 Chr.19: 10575026-10575026 on Build GRCh38
rs367834302 Chr.19: 10574949-10574949 on Build GRCh38
rs199818168 Chr.19: 10575018-10575018 on Build GRCh38
rs537168983 Chr.19: 10575008-10575008 on Build GRCh38
rs375010828 Chr.19: 10575029-10575029 on Build GRCh38
rs756203484 Chr.19: 10574918-10574918 on Build GRCh38
rs200574564 Chr.19: 10574931-10574931 on Build GRCh38
rs764847596 Chr.19: 10574961-10574961 on Build GRCh38
rs536244443 Chr.19: 10574865-10574865 on Build GRCh38
rs772046920 Chr.19: 10574953-10574953 on Build GRCh38
rs943226399 Chr.19: 10574842-10574842 on Build GRCh38
rs373760173 Chr.19: 10574919-10574919 on Build GRCh38
rs545243621 Chr.19: 10575039-10575039 on Build GRCh38
rs755478860 Chr.19: 10574886-10574886 on Build GRCh38
rs368878874 Chr.19: 10574864-10574864 on Build GRCh38
rs201504302 Chr.19: 10574956-10574956 on Build GRCh38
rs751892630 Chr.19: 10574879-10574879 on Build GRCh38
rs760247029 Chr.19: 10574860-10574860 on Build GRCh38
rs867815531 Chr.19: 10574901-10574901 on Build GRCh38
rs370943914 Chr.19: 10574876-10574876 on Build GRCh38
rs746389036 Chr.19: 10574945-10574945 on Build GRCh38
rs759874264 Chr.19: 10574967-10574967 on Build GRCh38
rs959961310 Chr.19: 10574928-10574928 on Build GRCh38
rs761378346 Chr.19: 10574873-10574873 on Build GRCh38
rs764217218 Chr.19: 10575014-10575014 on Build GRCh38
rs776152265 Chr.19: 10574958-10574958 on Build GRCh38
rs897242026 Chr.19: 10574832-10574832 on Build GRCh38
rs761478533 Chr.19: 10574960-10574960 on Build GRCh38
rs17849899 Chr.19: 10574987-10574987 on Build GRCh38
rs968875193 Chr.19: 10574963-10574963 on Build GRCh38
rs753185749 Chr.19: 10575007-10575007 on Build GRCh38
rs555968863 Chr.19: 10574942-10574942 on Build GRCh38
rs905866788 Chr.19: 10574874-10574874 on Build GRCh38

View Full Product Details