Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132729659-132729916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKLE2
Entrez Gene ID
23141
Gene Name
ankyrin repeat and LEM domain containing 2
Gene Aliases
KIAA0692, LEMD7, Lem4, MCPH16
UniGene
Hs.654628
Gene Chromosome Location
Chr.12: 132725667-132761888 on Build GRCh38
Transcripts
NM_015114.2 XM_005266159.3 XM_005266160.1 XM_006719735.1 XM_017019076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4979701 c.2472C>G p.L824L Chr.12: 132729690-132729690 on Build GRCh38
1188668 c.2366G>A p.R789K Chr.12: 132729796-132729796 on Build GRCh38
4979702 c.2333C>G p.S778C Chr.12: 132729829-132729829 on Build GRCh38
7080169 c.2297G>A p.R766K Chr.12: 132729865-132729865 on Build GRCh38
7101193 c.2367G>A p.R789R Chr.12: 132729795-132729795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369320391 Chr.12: 132729679-132729679 on Build GRCh38
rs759717835 Chr.12: 132729750-132729750 on Build GRCh38
rs1037509093 Chr.12: 132729693-132729693 on Build GRCh38
rs186469877 Chr.12: 132729824-132729824 on Build GRCh38
rs755196191 Chr.12: 132729699-132729699 on Build GRCh38
rs773697339 Chr.12: 132729733-132729733 on Build GRCh38
rs750134734 Chr.12: 132729845-132729845 on Build GRCh38
rs376530487 Chr.12: 132729812-132729812 on Build GRCh38
rs572111545 Chr.12: 132729753-132729753 on Build GRCh38
rs754976405 Chr.12: 132729860-132729860 on Build GRCh38
rs753976276 Chr.12: 132729695-132729695 on Build GRCh38
rs369591537 Chr.12: 132729796-132729796 on Build GRCh38
rs761024797 Chr.12: 132729740-132729740 on Build GRCh38
rs201785518 Chr.12: 132729818-132729818 on Build GRCh38
rs530881578 Chr.12: 132729768-132729768 on Build GRCh38
rs200721415 Chr.12: 132729813-132729813 on Build GRCh38
rs753763983 Chr.12: 132729859-132729859 on Build GRCh38
rs754464109 Chr.12: 132729786-132729786 on Build GRCh38
rs776806854 Chr.12: 132729749-132729749 on Build GRCh38
rs559941180 Chr.12: 132729678-132729678 on Build GRCh38
rs749265166 Chr.12: 132729782-132729782 on Build GRCh38
rs745775918 Chr.12: 132729709-132729709 on Build GRCh38
rs748808201 Chr.12: 132729721-132729721 on Build GRCh38
rs565063145 Chr.12: 132729754-132729754 on Build GRCh38
rs200454083 Chr.12: 132729704-132729704 on Build GRCh38
rs375841027 Chr.12: 132729702-132729702 on Build GRCh38
rs766368315 Chr.12: 132729694-132729694 on Build GRCh38
rs779987897 Chr.12: 132729772-132729772 on Build GRCh38
rs376360110 Chr.12: 132729767-132729767 on Build GRCh38
rs768247207 Chr.12: 132729728-132729728 on Build GRCh38
rs1000297366 Chr.12: 132729737-132729737 on Build GRCh38
rs752269208 Chr.12: 132729876-132729876 on Build GRCh38
rs528690169 Chr.12: 132729703-132729703 on Build GRCh38
rs12809719 Chr.12: 132729722-132729722 on Build GRCh38
rs373826499 Chr.12: 132729848-132729848 on Build GRCh38
rs200992817 Chr.12: 132729825-132729825 on Build GRCh38
rs771764666 Chr.12: 132729800-132729800 on Build GRCh38
rs775703008 Chr.12: 132729823-132729823 on Build GRCh38
rs766212388 Chr.12: 132729853-132729853 on Build GRCh38
rs542277356 Chr.12: 132729843-132729843 on Build GRCh38
rs373384820 Chr.12: 132729719-132729719 on Build GRCh38
rs200179300 Chr.12: 132729815-132729815 on Build GRCh38
rs779472244 Chr.12: 132729720-132729720 on Build GRCh38
rs772312045 Chr.12: 132729732-132729732 on Build GRCh38
rs963399121 Chr.12: 132729847-132729847 on Build GRCh38
rs767008737 Chr.12: 132729741-132729741 on Build GRCh38
rs371526496 Chr.12: 132729681-132729681 on Build GRCh38
rs545538645 Chr.12: 132729708-132729708 on Build GRCh38
rs773963548 Chr.12: 132729731-132729731 on Build GRCh38
rs372987812 Chr.12: 132729805-132729805 on Build GRCh38
rs776299184 Chr.12: 132729828-132729830 on Build GRCh38
rs757940974 Chr.12: 132729880-132729880 on Build GRCh38
rs758700836 Chr.12: 132729755-132729755 on Build GRCh38
rs747751622 Chr.12: 132729798-132729798 on Build GRCh38
rs767211770 Chr.12: 132729759-132729759 on Build GRCh38
rs778331687 Chr.12: 132729870-132729870 on Build GRCh38

View Full Product Details