Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71732979-71733238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G1
Entrez Gene ID
164
Gene Name
adaptor related protein complex 1 gamma 1 subunit
Gene Aliases
ADTG, CLAPG1
UniGene
Hs.461253
Gene Chromosome Location
Chr.16: 71729000-71809073 on Build GRCh38
Transcripts
NM_001030007.1 NM_001128.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4784355 c.2393G>A p.R798Q Chr.16: 71733143-71733143 on Build GRCh38
6307614 c.2422C>A p.Q808K Chr.16: 71733105-71733105 on Build GRCh38
4784354 c.2453G>A p.R818Q Chr.16: 71733143-71733143 on Build GRCh38
5654318 c.2375T>G p.L792R Chr.16: 71733152-71733152 on Build GRCh38
6307613 c.2431C>A p.Q811K Chr.16: 71733105-71733105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056905240 Chr.16: 71733046-71733046 on Build GRCh38
rs768203007 Chr.16: 71733047-71733047 on Build GRCh38
rs749801429 Chr.16: 71733172-71733172 on Build GRCh38
rs768442011 Chr.16: 71733032-71733033 on Build GRCh38
rs147519448 Chr.16: 71733061-71733061 on Build GRCh38
rs899085166 Chr.16: 71733118-71733118 on Build GRCh38
rs764166650 Chr.16: 71733126-71733126 on Build GRCh38
rs118111752 Chr.16: 71733162-71733162 on Build GRCh38
rs780767852 Chr.16: 71733169-71733169 on Build GRCh38
rs768058576 Chr.16: 71733175-71733175 on Build GRCh38
rs781693456 Chr.16: 71733139-71733139 on Build GRCh38
rs753761796 Chr.16: 71733100-71733100 on Build GRCh38
rs1021860243 Chr.16: 71733091-71733091 on Build GRCh38
rs761467938 Chr.16: 71733057-71733057 on Build GRCh38
rs763008013 Chr.16: 71733115-71733115 on Build GRCh38
rs368796987 Chr.16: 71733109-71733109 on Build GRCh38
rs747784229 Chr.16: 71733184-71733184 on Build GRCh38
rs375099704 Chr.16: 71733199-71733199 on Build GRCh38
rs766353180 Chr.16: 71733099-71733099 on Build GRCh38
rs773944366 Chr.16: 71733053-71733053 on Build GRCh38
rs781714640 Chr.16: 71733143-71733143 on Build GRCh38
rs527422540 Chr.16: 71733075-71733075 on Build GRCh38
rs778435079 Chr.16: 71733178-71733178 on Build GRCh38
rs771747624 Chr.16: 71733189-71733189 on Build GRCh38
rs772947407 Chr.16: 71733070-71733070 on Build GRCh38
rs756528630 Chr.16: 71733168-71733168 on Build GRCh38
rs200448545 Chr.16: 71733031-71733031 on Build GRCh38
rs757454113 Chr.16: 71733142-71733142 on Build GRCh38
rs1012270333 Chr.16: 71733067-71733067 on Build GRCh38

View Full Product Details