Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74561990-74562251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
M1AP
Entrez Gene ID
130951
Gene Name
meiosis 1 associated protein
Gene Aliases
C2orf65, D6Mm5e, SPATA37
UniGene
Hs.348645
Gene Chromosome Location
Chr.2: 74557640-74648338 on Build GRCh38
Transcripts
NM_001281296.1 NM_001321739.1 NM_138804.4 XM_005264152.3 XM_006711946.3 XM_011532548.2 XM_011532549.2 XM_011532550.2 XM_011532551.2 XM_011532552.2 XM_011532553.2 XM_011532554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3739927 c.1281+6T>C p.? Chr.2: 74562211-74562211 on Build GRCh38
4095832 c.1256A>C p.H419P Chr.2: 74562242-74562242 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs564753623 Chr.2: 74562067-74562067 on Build GRCh38
rs752189876 Chr.2: 74562180-74562180 on Build GRCh38
rs781120910 Chr.2: 74562223-74562223 on Build GRCh38
rs941342785 Chr.2: 74562134-74562134 on Build GRCh38
rs909775752 Chr.2: 74562120-74562120 on Build GRCh38
rs73949685 Chr.2: 74562103-74562103 on Build GRCh38
rs772004777 Chr.2: 74562224-74562226 on Build GRCh38
rs777102056 Chr.2: 74562177-74562177 on Build GRCh38
rs143680043 Chr.2: 74562222-74562222 on Build GRCh38
rs889098780 Chr.2: 74562207-74562207 on Build GRCh38
rs922466937 Chr.2: 74562039-74562039 on Build GRCh38
rs547295213 Chr.2: 74562121-74562121 on Build GRCh38
rs761062952 Chr.2: 74562172-74562172 on Build GRCh38
rs1050839071 Chr.2: 74562204-74562204 on Build GRCh38
rs745662064 Chr.2: 74562172-74562172 on Build GRCh38
rs377032064 Chr.2: 74562202-74562202 on Build GRCh38
rs79405350 Chr.2: 74562175-74562175 on Build GRCh38
rs765800421 Chr.2: 74562178-74562178 on Build GRCh38
rs549744223 Chr.2: 74562019-74562019 on Build GRCh38
rs932460277 Chr.2: 74562076-74562076 on Build GRCh38
rs940805826 Chr.2: 74562107-74562107 on Build GRCh38

View Full Product Details