Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201704445-201704700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2
Entrez Gene ID
57679
Gene Name
ALS2, alsin Rho guanine nucleotide exchange factor
Gene Aliases
ALS2CR6, ALSJ, IAHSP, PLSJ
UniGene
Hs.471096
Gene Chromosome Location
Chr.2: 201700263-201781172 on Build GRCh38
Transcripts
NM_020919.3 XM_006712654.2 XM_006712655.3 XM_011511530.2 XM_017004569.1 XM_017004571.1 XM_017004572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3838304 c.4743G>A p.E1581E Chr.2: 201704549-201704549 on Build GRCh38
4090253 c.4831C>T p.R1611W Chr.2: 201704461-201704461 on Build GRCh38
3576370 c.4725C>T p.I1575I Chr.2: 201704567-201704567 on Build GRCh38
6570571 c.4757T>C p.V1586A Chr.2: 201704535-201704535 on Build GRCh38
4589748 c.4704_4705insC p.S1569fs*5 Chr.2: 201704587-201704588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772452966 Chr.2: 201704475-201704475 on Build GRCh38
rs369015911 Chr.2: 201704519-201704519 on Build GRCh38
rs777070358 Chr.2: 201704518-201704518 on Build GRCh38
rs1027849655 Chr.2: 201704626-201704626 on Build GRCh38
rs778530412 Chr.2: 201704635-201704635 on Build GRCh38
rs370296035 Chr.2: 201704472-201704472 on Build GRCh38
rs780039812 Chr.2: 201704617-201704617 on Build GRCh38
rs544314801 Chr.2: 201704663-201704663 on Build GRCh38
rs768514971 Chr.2: 201704636-201704636 on Build GRCh38
rs755994384 Chr.2: 201704540-201704541 on Build GRCh38
rs386134188 Chr.2: 201704571-201704571 on Build GRCh38
rs35110478 Chr.2: 201704528-201704528 on Build GRCh38
rs755629995 Chr.2: 201704613-201704613 on Build GRCh38
rs565284119 Chr.2: 201704591-201704591 on Build GRCh38
rs532630245 Chr.2: 201704600-201704600 on Build GRCh38
rs753780738 Chr.2: 201704632-201704632 on Build GRCh38
rs754768570 Chr.2: 201704633-201704633 on Build GRCh38
rs760849143 Chr.2: 201704507-201704507 on Build GRCh38
rs776508125 Chr.2: 201704651-201704651 on Build GRCh38
rs192324626 Chr.2: 201704664-201704664 on Build GRCh38
rs540726634 Chr.2: 201704529-201704529 on Build GRCh38
rs771402858 Chr.2: 201704514-201704514 on Build GRCh38
rs377417529 Chr.2: 201704540-201704540 on Build GRCh38
rs555220239 Chr.2: 201704474-201704474 on Build GRCh38
rs773373317 Chr.2: 201704498-201704498 on Build GRCh38

View Full Product Details