Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45213208-45213463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L2
Entrez Gene ID
90332
Gene Name
exocyst complex component 3 like 2
Gene Aliases
XTP7
UniGene
Hs.337557
Gene Chromosome Location
Chr.19: 45212621-45234211 on Build GRCh38
Transcripts
NM_138568.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4911091 c.1028T>C p.L343P Chr.19: 45213271-45213271 on Build GRCh38
7300314 c.998G>A p.R333H Chr.19: 45213301-45213301 on Build GRCh38
3692872 c.994G>A p.A332T Chr.19: 45213305-45213305 on Build GRCh38
3535865 c.1022G>A p.R341Q Chr.19: 45213277-45213277 on Build GRCh38
258778 c.974G>A p.R325H Chr.19: 45213325-45213325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372471842 Chr.19: 45213284-45213284 on Build GRCh38
rs530663749 Chr.19: 45213263-45213263 on Build GRCh38
rs747050393 Chr.19: 45213327-45213332 on Build GRCh38
rs933512365 Chr.19: 45213369-45213369 on Build GRCh38
rs781172895 Chr.19: 45213364-45213364 on Build GRCh38
rs747576250 Chr.19: 45213407-45213407 on Build GRCh38
rs923325123 Chr.19: 45213333-45213333 on Build GRCh38
rs192143624 Chr.19: 45213385-45213385 on Build GRCh38
rs374946353 Chr.19: 45213251-45213251 on Build GRCh38
rs375095520 Chr.19: 45213285-45213285 on Build GRCh38
rs750772950 Chr.19: 45213236-45213236 on Build GRCh38
rs367776193 Chr.19: 45213229-45213229 on Build GRCh38
rs780869435 Chr.19: 45213402-45213402 on Build GRCh38
rs756258972 Chr.19: 45213332-45213332 on Build GRCh38
rs773805836 Chr.19: 45213301-45213301 on Build GRCh38
rs755095883 Chr.19: 45213242-45213242 on Build GRCh38
rs765082833 Chr.19: 45213368-45213368 on Build GRCh38
rs1007090933 Chr.19: 45213318-45213318 on Build GRCh38
rs565176587 Chr.19: 45213382-45213382 on Build GRCh38
rs536261489 Chr.19: 45213240-45213240 on Build GRCh38
rs547412844 Chr.19: 45213377-45213377 on Build GRCh38
rs769409244 Chr.19: 45213409-45213409 on Build GRCh38
rs758075637 Chr.19: 45213380-45213380 on Build GRCh38
rs148132329 Chr.19: 45213291-45213291 on Build GRCh38
rs747735507 Chr.19: 45213429-45213429 on Build GRCh38
rs573616397 Chr.19: 45213235-45213235 on Build GRCh38
rs371350226 Chr.19: 45213239-45213239 on Build GRCh38
rs346740 Chr.19: 45213420-45213420 on Build GRCh38
rs771973379 Chr.19: 45213278-45213278 on Build GRCh38
rs751772044 Chr.19: 45213309-45213309 on Build GRCh38
rs368051115 Chr.19: 45213279-45213279 on Build GRCh38
rs545279176 Chr.19: 45213317-45213317 on Build GRCh38
rs762329905 Chr.19: 45213290-45213290 on Build GRCh38
rs763420511 Chr.19: 45213238-45213238 on Build GRCh38
rs901308464 Chr.19: 45213384-45213384 on Build GRCh38
rs762315844 Chr.19: 45213230-45213230 on Build GRCh38
rs1025989685 Chr.19: 45213423-45213423 on Build GRCh38
rs201048859 Chr.19: 45213272-45213272 on Build GRCh38
rs754644497 Chr.19: 45213400-45213400 on Build GRCh38
rs369915015 Chr.19: 45213306-45213306 on Build GRCh38
rs745620424 Chr.19: 45213269-45213269 on Build GRCh38
rs770174254 Chr.19: 45213297-45213297 on Build GRCh38
rs778960279 Chr.19: 45213266-45213266 on Build GRCh38
rs781433775 Chr.19: 45213247-45213247 on Build GRCh38
rs759841252 Chr.19: 45213316-45213316 on Build GRCh38
rs776146386 Chr.19: 45213287-45213287 on Build GRCh38
rs763222027 Chr.19: 45213302-45213302 on Build GRCh38
rs764606029 Chr.19: 45213438-45213438 on Build GRCh38
rs753760745 Chr.19: 45213250-45213250 on Build GRCh38
rs752805504 Chr.19: 45213325-45213325 on Build GRCh38

View Full Product Details