Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149200271-149200544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM3
Entrez Gene ID
22885
Gene Name
actin binding LIM protein family member 3
Gene Aliases
HMFN1661
UniGene
Hs.49688
Gene Chromosome Location
Chr.5: 149141483-149260439 on Build GRCh38
Transcripts
NM_001301015.1 NM_001301018.1 NM_001301027.1 NM_001301028.1 NM_014945.3 XM_005268392.1 XM_005268393.1 XM_005268394.1 XM_005268395.1 XM_005268396.1 XM_011537609.2 XM_017009238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268555 c.430A>G p.K144E Chr.5: 149200410-149200410 on Build GRCh38
5574373 c.439G>C p.G147R Chr.5: 149200419-149200419 on Build GRCh38
3661591 c.440G>T p.G147V Chr.5: 149200420-149200420 on Build GRCh38
3661589 c.439G>T p.G147* Chr.5: 149200419-149200419 on Build GRCh38
232609 c.341C>T p.P114L Chr.5: 149200321-149200321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751536510 Chr.5: 149200470-149200470 on Build GRCh38
rs770047907 Chr.5: 149200313-149200313 on Build GRCh38
rs776400534 Chr.5: 149200305-149200305 on Build GRCh38
rs750132789 Chr.5: 149200453-149200453 on Build GRCh38
rs35907283 Chr.5: 149200354-149200354 on Build GRCh38
rs944628805 Chr.5: 149200389-149200389 on Build GRCh38
rs527956302 Chr.5: 149200385-149200385 on Build GRCh38
rs114255499 Chr.5: 149200346-149200346 on Build GRCh38
rs10055838 Chr.5: 149200376-149200376 on Build GRCh38
rs376272159 Chr.5: 149200443-149200443 on Build GRCh38
rs761953405 Chr.5: 149200381-149200381 on Build GRCh38
rs1048741274 Chr.5: 149200345-149200345 on Build GRCh38
rs960455769 Chr.5: 149200493-149200493 on Build GRCh38
rs773828506 Chr.5: 149200322-149200322 on Build GRCh38
rs41291951 Chr.5: 149200497-149200497 on Build GRCh38
rs1021667720 Chr.5: 149200444-149200444 on Build GRCh38
rs746685732 Chr.5: 149200302-149200302 on Build GRCh38
rs878871139 Chr.5: 149200418-149200418 on Build GRCh38
rs774821112 Chr.5: 149200417-149200417 on Build GRCh38
rs376511147 Chr.5: 149200379-149200379 on Build GRCh38
rs749832445 Chr.5: 149200405-149200405 on Build GRCh38
rs748584807 Chr.5: 149200311-149200311 on Build GRCh38
rs754592227 Chr.5: 149200472-149200472 on Build GRCh38
rs1023600295 Chr.5: 149200512-149200512 on Build GRCh38
rs562950851 Chr.5: 149200352-149200352 on Build GRCh38
rs758336879 Chr.5: 149200442-149200442 on Build GRCh38
rs759655987 Chr.5: 149200420-149200420 on Build GRCh38
rs373339694 Chr.5: 149200353-149200353 on Build GRCh38
rs568903305 Chr.5: 149200523-149200523 on Build GRCh38
rs762785799 Chr.5: 149200460-149200460 on Build GRCh38
rs770209379 Chr.5: 149200392-149200392 on Build GRCh38
rs778782219 Chr.5: 149200304-149200304 on Build GRCh38
rs370614667 Chr.5: 149200397-149200397 on Build GRCh38
rs41291949 Chr.5: 149200489-149200489 on Build GRCh38
rs967685531 Chr.5: 149200416-149200416 on Build GRCh38
rs774374268 Chr.5: 149200330-149200330 on Build GRCh38
rs752401813 Chr.5: 149200480-149200480 on Build GRCh38
rs780792369 Chr.5: 149200479-149200479 on Build GRCh38
rs6580602 Chr.5: 149200462-149200462 on Build GRCh38
rs76939309 Chr.5: 149200412-149200412 on Build GRCh38
rs758224799 Chr.5: 149200375-149200375 on Build GRCh38
rs143160468 Chr.5: 149200339-149200339 on Build GRCh38
rs977807733 Chr.5: 149200428-149200428 on Build GRCh38
rs779636155 Chr.5: 149200301-149200301 on Build GRCh38
rs772137973 Chr.5: 149200429-149200429 on Build GRCh38
rs750375674 Chr.5: 149200366-149200366 on Build GRCh38
rs149111210 Chr.5: 149200329-149200329 on Build GRCh38

View Full Product Details