Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48454909-48455162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP22
Entrez Gene ID
58504
Gene Name
Rho GTPase activating protein 22
Gene Aliases
RhoGAP2, RhoGap22
UniGene
Hs.655672
Gene Chromosome Location
Chr.10: 48429831-48656265 on Build GRCh38
Transcripts
NM_001256024.1 NM_001256025.2 NM_001256026.1 NM_021226.3 NR_045675.1 XM_005270014.3 XM_011540002.1 XM_011540003.1 XM_011540004.1 XM_011540005.1 XM_011540006.1 XM_011540007.2 XM_011540011.2 XM_011540012.2 XM_011540013.2 XM_011540015.2 XM_017016470.1 XM_017016471.1 XM_017016472.1 XM_017016473.1 XM_017016474.1 XM_017016475.1 XM_017016476.1 XM_017016477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5359787 c.390-5delC p.? Chr.10: 48455139-48455139 on Build GRCh38
4558115 c.437G>A p.R146Q Chr.10: 48455087-48455087 on Build GRCh38
3867292 c.788G>A p.R263K Chr.10: 48455054-48455054 on Build GRCh38
5370697 c.769C>T p.P257S Chr.10: 48455073-48455073 on Build GRCh38
6435451 c.832G>A p.E278K Chr.10: 48455010-48455010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752931097 Chr.10: 48454980-48454980 on Build GRCh38
rs76158077 Chr.10: 48455125-48455125 on Build GRCh38
rs751767741 Chr.10: 48455050-48455050 on Build GRCh38
rs202246605 Chr.10: 48455139-48455139 on Build GRCh38
rs780756039 Chr.10: 48455085-48455085 on Build GRCh38
rs777081814 Chr.10: 48455005-48455005 on Build GRCh38
rs749140905 Chr.10: 48454954-48454954 on Build GRCh38
rs757843608 Chr.10: 48455075-48455075 on Build GRCh38
rs765033364 Chr.10: 48455129-48455129 on Build GRCh38
rs780893219 Chr.10: 48455142-48455142 on Build GRCh38
rs779541553 Chr.10: 48455076-48455076 on Build GRCh38
rs770347078 Chr.10: 48455088-48455088 on Build GRCh38
rs569942959 Chr.10: 48454942-48454942 on Build GRCh38
rs72796308 Chr.10: 48455141-48455141 on Build GRCh38
rs753645581 Chr.10: 48455059-48455059 on Build GRCh38
rs35021101 Chr.10: 48455062-48455062 on Build GRCh38
rs774595774 Chr.10: 48455115-48455115 on Build GRCh38
rs763561069 Chr.10: 48455099-48455099 on Build GRCh38
rs769648611 Chr.10: 48455138-48455139 on Build GRCh38
rs750290756 Chr.10: 48455137-48455137 on Build GRCh38
rs759834785 Chr.10: 48455054-48455054 on Build GRCh38
rs573657113 Chr.10: 48455010-48455010 on Build GRCh38
rs758618807 Chr.10: 48455096-48455096 on Build GRCh38
rs201247202 Chr.10: 48454996-48454996 on Build GRCh38
rs571150652 Chr.10: 48455071-48455071 on Build GRCh38
rs759584335 Chr.10: 48455116-48455116 on Build GRCh38
rs773894826 Chr.10: 48455032-48455032 on Build GRCh38
rs141947668 Chr.10: 48455077-48455077 on Build GRCh38
rs975964307 Chr.10: 48454955-48454955 on Build GRCh38
rs763119711 Chr.10: 48455049-48455049 on Build GRCh38
rs745354859 Chr.10: 48454967-48454967 on Build GRCh38
rs1029588167 Chr.10: 48455132-48455132 on Build GRCh38
rs989981152 Chr.10: 48455018-48455018 on Build GRCh38
rs752933087 Chr.10: 48455124-48455124 on Build GRCh38
rs773839704 Chr.10: 48455095-48455095 on Build GRCh38
rs537319585 Chr.10: 48454991-48454991 on Build GRCh38
rs140608042 Chr.10: 48455067-48455067 on Build GRCh38
rs74800997 Chr.10: 48454989-48454989 on Build GRCh38
rs748513396 Chr.10: 48455087-48455087 on Build GRCh38
rs772863523 Chr.10: 48454947-48454947 on Build GRCh38
rs778875323 Chr.10: 48454966-48454966 on Build GRCh38
rs757241600 Chr.10: 48454965-48454965 on Build GRCh38
rs762173518 Chr.10: 48455011-48455011 on Build GRCh38
rs534705917 Chr.10: 48455031-48455031 on Build GRCh38
rs779486546 Chr.10: 48455140-48455140 on Build GRCh38
rs563448239 Chr.10: 48455104-48455104 on Build GRCh38
rs771640977 Chr.10: 48454987-48454987 on Build GRCh38
rs760644302 Chr.10: 48455117-48455117 on Build GRCh38
rs763749142 Chr.10: 48455058-48455058 on Build GRCh38

View Full Product Details