Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7707637-7707898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY2
Entrez Gene ID
108
Gene Name
adenylate cyclase 2
Gene Aliases
AC2, HBAC2
UniGene
Hs.481545
Gene Chromosome Location
Chr.5: 7396230-7830081 on Build GRCh38
Transcripts
NM_020546.2 XM_011513942.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671496 c.826C>T p.H276Y Chr.5: 7707803-7707803 on Build GRCh38
4625606 c.788A>C p.E263A Chr.5: 7707765-7707765 on Build GRCh38
1671495 c.1366C>T p.H456Y Chr.5: 7707803-7707803 on Build GRCh38
739186 c.1401G>T p.K467N Chr.5: 7707838-7707838 on Build GRCh38
4956690 c.791G>A p.G264E Chr.5: 7707768-7707768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765658507 Chr.5: 7707787-7707787 on Build GRCh38
rs961765370 Chr.5: 7707853-7707854 on Build GRCh38
rs140842335 Chr.5: 7707749-7707749 on Build GRCh38
rs760207405 Chr.5: 7707779-7707779 on Build GRCh38
rs747410725 Chr.5: 7707809-7707810 on Build GRCh38
rs757036389 Chr.5: 7707863-7707863 on Build GRCh38
rs763559291 Chr.5: 7707800-7707800 on Build GRCh38
rs149736529 Chr.5: 7707754-7707754 on Build GRCh38
rs112493968 Chr.5: 7707748-7707748 on Build GRCh38
rs547704282 Chr.5: 7707856-7707856 on Build GRCh38
rs745777282 Chr.5: 7707868-7707868 on Build GRCh38
rs752098719 Chr.5: 7707808-7707808 on Build GRCh38
rs748295491 Chr.5: 7707694-7707694 on Build GRCh38
rs147852844 Chr.5: 7707778-7707778 on Build GRCh38
rs1030943666 Chr.5: 7707671-7707671 on Build GRCh38
rs781532749 Chr.5: 7707690-7707690 on Build GRCh38
rs767463191 Chr.5: 7707841-7707841 on Build GRCh38
rs142609557 Chr.5: 7707746-7707746 on Build GRCh38
rs372469100 Chr.5: 7707853-7707853 on Build GRCh38
rs777389360 Chr.5: 7707736-7707736 on Build GRCh38
rs752992620 Chr.5: 7707847-7707847 on Build GRCh38
rs376018386 Chr.5: 7707867-7707867 on Build GRCh38
rs749266162 Chr.5: 7707739-7707739 on Build GRCh38
rs773392259 Chr.5: 7707790-7707790 on Build GRCh38
rs34899441 Chr.5: 7707805-7707805 on Build GRCh38
rs755974474 Chr.5: 7707707-7707707 on Build GRCh38
rs972707000 Chr.5: 7707860-7707860 on Build GRCh38
rs200793395 Chr.5: 7707783-7707783 on Build GRCh38

View Full Product Details