Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134928981-134929254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH8A1
Entrez Gene ID
64577
Gene Name
aldehyde dehydrogenase 8 family member A1
Gene Aliases
ALDH12, DJ352A20.2
UniGene
Hs.486520
Gene Chromosome Location
Chr.6: 134917390-134950122 on Build GRCh38
Transcripts
NM_001193480.1 NM_022568.3 NM_170771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4519356 c.1006G>A p.E336K Chr.6: 134929059-134929059 on Build GRCh38
234378 c.939G>A p.K313K Chr.6: 134929126-134929126 on Build GRCh38
6105098 c.982G>T p.A328S Chr.6: 134929083-134929083 on Build GRCh38
1244821 c.1010A>C p.K337T Chr.6: 134929055-134929055 on Build GRCh38
4661059 c.948C>T p.V316V Chr.6: 134929117-134929117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368484988 Chr.6: 134929102-134929102 on Build GRCh38
rs56825243 Chr.6: 134929098-134929098 on Build GRCh38
rs778416593 Chr.6: 134929207-134929207 on Build GRCh38
rs374129470 Chr.6: 134929022-134929022 on Build GRCh38
rs375341491 Chr.6: 134929145-134929145 on Build GRCh38
rs149971112 Chr.6: 134929165-134929165 on Build GRCh38
rs201021285 Chr.6: 134929171-134929171 on Build GRCh38
rs749735779 Chr.6: 134929047-134929047 on Build GRCh38
rs772291428 Chr.6: 134929082-134929082 on Build GRCh38
rs756386224 Chr.6: 134929042-134929042 on Build GRCh38
rs776682957 Chr.6: 134929099-134929099 on Build GRCh38
rs1027686847 Chr.6: 134929226-134929226 on Build GRCh38
rs772855340 Chr.6: 134929104-134929104 on Build GRCh38
rs745879607 Chr.6: 134929081-134929081 on Build GRCh38
rs762496740 Chr.6: 134929105-134929105 on Build GRCh38
rs376511348 Chr.6: 134929101-134929101 on Build GRCh38
rs746847645 Chr.6: 134929090-134929090 on Build GRCh38
rs931546292 Chr.6: 134929232-134929232 on Build GRCh38
rs959071003 Chr.6: 134929234-134929234 on Build GRCh38
rs758428952 Chr.6: 134929169-134929169 on Build GRCh38
rs757717892 Chr.6: 134929116-134929116 on Build GRCh38
rs761430844 Chr.6: 134929013-134929013 on Build GRCh38
rs897791244 Chr.6: 134929161-134929161 on Build GRCh38
rs200559431 Chr.6: 134929010-134929010 on Build GRCh38
rs762584218 Chr.6: 134929229-134929229 on Build GRCh38
rs766065061 Chr.6: 134929035-134929035 on Build GRCh38
rs368713329 Chr.6: 134929193-134929193 on Build GRCh38
rs372089376 Chr.6: 134929117-134929117 on Build GRCh38
rs943832809 Chr.6: 134929138-134929138 on Build GRCh38
rs747887113 Chr.6: 134929213-134929213 on Build GRCh38
rs370245282 Chr.6: 134929054-134929054 on Build GRCh38
rs761150774 Chr.6: 134929122-134929122 on Build GRCh38
rs768302068 Chr.6: 134929166-134929166 on Build GRCh38
rs764822240 Chr.6: 134929018-134929018 on Build GRCh38
rs367637050 Chr.6: 134929023-134929023 on Build GRCh38
rs1033708119 Chr.6: 134929014-134929014 on Build GRCh38
rs374461989 Chr.6: 134929087-134929087 on Build GRCh38
rs373345480 Chr.6: 134929204-134929204 on Build GRCh38
rs563171209 Chr.6: 134929129-134929129 on Build GRCh38
rs769868127 Chr.6: 134929222-134929222 on Build GRCh38
rs776388005 Chr.6: 134929012-134929012 on Build GRCh38
rs773236195 Chr.6: 134929225-134929225 on Build GRCh38
rs868393412 Chr.6: 134929115-134929115 on Build GRCh38
rs377167242 Chr.6: 134929205-134929205 on Build GRCh38
rs751133124 Chr.6: 134929118-134929118 on Build GRCh38
rs542548050 Chr.6: 134929227-134929227 on Build GRCh38
rs770317085 Chr.6: 134929231-134929231 on Build GRCh38
rs753121308 Chr.6: 134929041-134929041 on Build GRCh38
rs754433783 Chr.6: 134929132-134929132 on Build GRCh38
rs267600820 Chr.6: 134929214-134929214 on Build GRCh38
rs778959002 Chr.6: 134929064-134929064 on Build GRCh38
rs778375690 Chr.6: 134929043-134929043 on Build GRCh38

View Full Product Details