Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70729253-70729484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
-
UniGene
Hs.352156
Gene Chromosome Location
Chr.10: 70672518-70762441 on Build GRCh38
Transcripts
NM_080722.4 NM_139155.3 XM_011539300.2 XM_011539301.2 XM_011539302.2 XM_011539303.2 XM_011539305.2 XM_011539306.2 XM_011539307.2 XM_011539308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1348855 c.925G>A p.V309I Chr.10: 70729348-70729348 on Build GRCh38
1348854 c.924C>T p.L308L Chr.10: 70729347-70729347 on Build GRCh38
4519032 c.921_922CC>TT p.L308F Chr.10: 70729344-70729345 on Build GRCh38
4513373 c.936C>T p.I312I Chr.10: 70729359-70729359 on Build GRCh38
5761351 c.950G>A p.R317Q Chr.10: 70729373-70729373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751299515 Chr.10: 70729334-70729334 on Build GRCh38
rs201922304 Chr.10: 70729352-70729352 on Build GRCh38
rs375416194 Chr.10: 70729409-70729409 on Build GRCh38
rs543657491 Chr.10: 70729348-70729348 on Build GRCh38
rs756083617 Chr.10: 70729351-70729351 on Build GRCh38
rs145632694 Chr.10: 70729328-70729328 on Build GRCh38
rs199886417 Chr.10: 70729372-70729372 on Build GRCh38
rs761021660 Chr.10: 70729312-70729312 on Build GRCh38
rs746010286 Chr.10: 70729292-70729292 on Build GRCh38
rs772351639 Chr.10: 70729307-70729307 on Build GRCh38
rs1012450714 Chr.10: 70729315-70729315 on Build GRCh38
rs956385257 Chr.10: 70729350-70729350 on Build GRCh38
rs777006942 Chr.10: 70729383-70729383 on Build GRCh38
rs143037838 Chr.10: 70729347-70729347 on Build GRCh38
rs977780279 Chr.10: 70729373-70729373 on Build GRCh38
rs766128304 Chr.10: 70729333-70729333 on Build GRCh38
rs185947794 Chr.10: 70729408-70729408 on Build GRCh38
rs754798907 Chr.10: 70729345-70729345 on Build GRCh38
rs140471498 Chr.10: 70729327-70729327 on Build GRCh38
rs200944211 Chr.10: 70729422-70729422 on Build GRCh38
rs773662072 Chr.10: 70729398-70729398 on Build GRCh38
rs755919948 Chr.10: 70729418-70729418 on Build GRCh38
rs768956701 Chr.10: 70729380-70729380 on Build GRCh38
rs927903412 Chr.10: 70729455-70729455 on Build GRCh38
rs374659925 Chr.10: 70729419-70729419 on Build GRCh38
rs780215705 Chr.10: 70729359-70729359 on Build GRCh38
rs141048259 Chr.10: 70729326-70729326 on Build GRCh38
rs374572428 Chr.10: 70729342-70729342 on Build GRCh38
rs750470451 Chr.10: 70729402-70729402 on Build GRCh38
rs758479463 Chr.10: 70729424-70729424 on Build GRCh38
rs933808086 Chr.10: 70729450-70729451 on Build GRCh38
rs143268347 Chr.10: 70729318-70729318 on Build GRCh38
rs138495173 Chr.10: 70729331-70729331 on Build GRCh38
rs548298435 Chr.10: 70729384-70729384 on Build GRCh38
rs145120858 Chr.10: 70729353-70729353 on Build GRCh38
rs770325748 Chr.10: 70729385-70729385 on Build GRCh38
rs764478735 Chr.10: 70729314-70729314 on Build GRCh38
rs570771760 Chr.10: 70729450-70729450 on Build GRCh38
rs759293064 Chr.10: 70729403-70729403 on Build GRCh38
rs35906753 Chr.10: 70729311-70729311 on Build GRCh38

View Full Product Details