Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74830275-74830512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX3
Entrez Gene ID
594855
Gene Name
complexin 3
Gene Aliases
CPX-III, CPXIII, Nbla11589
UniGene
Hs.187694
Gene Chromosome Location
Chr.15: 74826610-74831795 on Build GRCh38
Transcripts
NM_001030005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3504010 c.424C>T p.Q142* Chr.15: 74830301-74830301 on Build GRCh38
5896238 c.415G>A p.D139N Chr.15: 74830292-74830292 on Build GRCh38
1938881 c.426G>C p.Q142H Chr.15: 74830303-74830303 on Build GRCh38
5383834 c.414G>A p.K138K Chr.15: 74830291-74830291 on Build GRCh38
1938883 c.461A>G p.K154R Chr.15: 74830338-74830338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753894200 Chr.15: 74830346-74830346 on Build GRCh38
rs141104376 Chr.15: 74830466-74830466 on Build GRCh38
rs760087747 Chr.15: 74830380-74830380 on Build GRCh38
rs779982482 Chr.15: 74830345-74830345 on Build GRCh38
rs924826574 Chr.15: 74830406-74830406 on Build GRCh38
rs1049433493 Chr.15: 74830362-74830362 on Build GRCh38
rs751061419 Chr.15: 74830310-74830310 on Build GRCh38
rs896973887 Chr.15: 74830412-74830412 on Build GRCh38
rs766880854 Chr.15: 74830392-74830392 on Build GRCh38
rs760401104 Chr.15: 74830395-74830395 on Build GRCh38
rs747181678 Chr.15: 74830369-74830369 on Build GRCh38
rs775564497 Chr.15: 74830312-74830312 on Build GRCh38
rs775822421 Chr.15: 74830388-74830388 on Build GRCh38
rs372733021 Chr.15: 74830298-74830298 on Build GRCh38
rs199540758 Chr.15: 74830303-74830303 on Build GRCh38
rs750148517 Chr.15: 74830393-74830393 on Build GRCh38
rs771347048 Chr.15: 74830371-74830371 on Build GRCh38
rs1023603316 Chr.15: 74830440-74830440 on Build GRCh38
rs952633307 Chr.15: 74830433-74830433 on Build GRCh38
rs368430383 Chr.15: 74830365-74830365 on Build GRCh38
rs567043629 Chr.15: 74830435-74830435 on Build GRCh38
rs763638070 Chr.15: 74830308-74830308 on Build GRCh38
rs374967692 Chr.15: 74830364-74830364 on Build GRCh38
rs970761151 Chr.15: 74830449-74830449 on Build GRCh38
rs755073779 Chr.15: 74830367-74830367 on Build GRCh38
rs749978130 Chr.15: 74830333-74830333 on Build GRCh38
rs551795671 Chr.15: 74830419-74830419 on Build GRCh38
rs889512938 Chr.15: 74830366-74830366 on Build GRCh38
rs751854584 Chr.15: 74830323-74830323 on Build GRCh38
rs761392286 Chr.15: 74830389-74830389 on Build GRCh38
rs533626135 Chr.15: 74830376-74830376 on Build GRCh38
rs947000094 Chr.15: 74830361-74830361 on Build GRCh38
rs770310639 Chr.15: 74830387-74830387 on Build GRCh38
rs1026900616 Chr.15: 74830428-74830428 on Build GRCh38
rs562166313 Chr.15: 74830368-74830368 on Build GRCh38
rs867613978 Chr.15: 74830423-74830423 on Build GRCh38
rs982198182 Chr.15: 74830451-74830451 on Build GRCh38
rs761172588 Chr.15: 74830349-74830350 on Build GRCh38
rs1045510272 Chr.15: 74830429-74830429 on Build GRCh38
rs755952079 Chr.15: 74830339-74830339 on Build GRCh38

View Full Product Details