Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55151716-55151943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP5
Entrez Gene ID
9787
Gene Name
DLG associated protein 5
Gene Aliases
DLG7, HURP
UniGene
Hs.77695
Gene Chromosome Location
Chr.14: 55148116-55191678 on Build GRCh38
Transcripts
NM_001146015.1 NM_014750.4 XM_017021840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4627274 c.2138A>T p.D713V Chr.14: 55151925-55151925 on Build GRCh38
5101873 c.2226_2227insA p.E743fs*25 Chr.14: 55151836-55151837 on Build GRCh38
1300677 c.2182C>G p.L728V Chr.14: 55151881-55151881 on Build GRCh38
956351 c.2173A>G p.S725G Chr.14: 55151890-55151890 on Build GRCh38
3793738 c.2231G>C p.G744A Chr.14: 55151832-55151832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779897612 Chr.14: 55151815-55151815 on Build GRCh38
rs771230352 Chr.14: 55151865-55151865 on Build GRCh38
rs371872680 Chr.14: 55151859-55151859 on Build GRCh38
rs1046840948 Chr.14: 55151866-55151866 on Build GRCh38
rs1033371108 Chr.14: 55151842-55151842 on Build GRCh38
rs895075639 Chr.14: 55151853-55151853 on Build GRCh38
rs554990547 Chr.14: 55151827-55151827 on Build GRCh38
rs772765834 Chr.14: 55151762-55151762 on Build GRCh38
rs754169309 Chr.14: 55151796-55151796 on Build GRCh38
rs749828854 Chr.14: 55151763-55151763 on Build GRCh38
rs35954941 Chr.14: 55151806-55151806 on Build GRCh38
rs552731400 Chr.14: 55151783-55151783 on Build GRCh38
rs760794371 Chr.14: 55151900-55151900 on Build GRCh38
rs751999434 Chr.14: 55151745-55151745 on Build GRCh38
rs768214354 Chr.14: 55151847-55151847 on Build GRCh38
rs80066134 Chr.14: 55151761-55151761 on Build GRCh38
rs776265591 Chr.14: 55151850-55151850 on Build GRCh38
rs931170692 Chr.14: 55151818-55151818 on Build GRCh38
rs144439184 Chr.14: 55151893-55151893 on Build GRCh38
rs374978881 Chr.14: 55151799-55151799 on Build GRCh38
rs761462822 Chr.14: 55151863-55151863 on Build GRCh38
rs768035423 Chr.14: 55151880-55151880 on Build GRCh38
rs781305986 Chr.14: 55151778-55151780 on Build GRCh38
rs954964576 Chr.14: 55151757-55151757 on Build GRCh38
rs369636499 Chr.14: 55151903-55151903 on Build GRCh38
rs764335998 Chr.14: 55151901-55151901 on Build GRCh38
rs756184072 Chr.14: 55151787-55151787 on Build GRCh38
rs147229036 Chr.14: 55151800-55151800 on Build GRCh38
rs778114777 Chr.14: 55151791-55151791 on Build GRCh38
rs772079555 Chr.14: 55151812-55151812 on Build GRCh38
rs1054950091 Chr.14: 55151837-55151837 on Build GRCh38
rs759917369 Chr.14: 55151871-55151871 on Build GRCh38
rs746506146 Chr.14: 55151825-55151825 on Build GRCh38
rs748213680 Chr.14: 55151817-55151819 on Build GRCh38

View Full Product Details