Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36151319-36151580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7A1
Entrez Gene ID
1346
Gene Name
cytochrome c oxidase subunit 7A1
Gene Aliases
COX7A, COX7AH, COX7AM
UniGene
Hs.421621
Gene Chromosome Location
Chr.19: 36150922-36152869 on Build GRCh38
Transcripts
NM_001864.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5855567 c.119C>T p.P40L Chr.19: 36151530-36151530 on Build GRCh38
3199575 c.105_107delGGA p.E35delE Chr.19: 36151542-36151544 on Build GRCh38
3796992 c.171G>A p.M57I Chr.19: 36151478-36151478 on Build GRCh38
4077343 c.129G>A p.L43L Chr.19: 36151520-36151520 on Build GRCh38
6634358 c.173C>T p.T58M Chr.19: 36151476-36151476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760849616 Chr.19: 36151504-36151504 on Build GRCh38
rs1022370010 Chr.19: 36151392-36151392 on Build GRCh38
rs769604961 Chr.19: 36151557-36151557 on Build GRCh38
rs774812314 Chr.19: 36151559-36151559 on Build GRCh38
rs764691465 Chr.19: 36151412-36151412 on Build GRCh38
rs762420991 Chr.19: 36151413-36151413 on Build GRCh38
rs958591246 Chr.19: 36151549-36151549 on Build GRCh38
rs758307661 Chr.19: 36151529-36151529 on Build GRCh38
rs766507690 Chr.19: 36151509-36151509 on Build GRCh38
rs903284005 Chr.19: 36151462-36151462 on Build GRCh38
rs756846859 Chr.19: 36151543-36151543 on Build GRCh38
rs748021382 Chr.19: 36151436-36151436 on Build GRCh38
rs762933059 Chr.19: 36151512-36151512 on Build GRCh38
rs17882491 Chr.19: 36151519-36151519 on Build GRCh38
rs776409233 Chr.19: 36151468-36151468 on Build GRCh38
rs754118291 Chr.19: 36151514-36151514 on Build GRCh38
rs199837220 Chr.19: 36151427-36151427 on Build GRCh38
rs371411166 Chr.19: 36151434-36151434 on Build GRCh38
rs557992806 Chr.19: 36151538-36151538 on Build GRCh38
rs780576270 Chr.19: 36151548-36151548 on Build GRCh38
rs201471888 Chr.19: 36151432-36151432 on Build GRCh38
rs764976179 Chr.19: 36151517-36151517 on Build GRCh38
rs369479742 Chr.19: 36151470-36151470 on Build GRCh38
rs372980477 Chr.19: 36151495-36151495 on Build GRCh38
rs751337144 Chr.19: 36151554-36151555 on Build GRCh38
rs565829737 Chr.19: 36151431-36151431 on Build GRCh38
rs80050273 Chr.19: 36151447-36151447 on Build GRCh38
rs774863677 Chr.19: 36151471-36151471 on Build GRCh38
rs745444096 Chr.19: 36151553-36151553 on Build GRCh38
rs150190766 Chr.19: 36151488-36151488 on Build GRCh38
rs746036352 Chr.19: 36151378-36151378 on Build GRCh38
rs764734167 Chr.19: 36151495-36151495 on Build GRCh38
rs546090932 Chr.19: 36151355-36151355 on Build GRCh38
rs34667074 Chr.19: 36151559-36151560 on Build GRCh38
rs754836489 Chr.19: 36151435-36151435 on Build GRCh38
rs375222289 Chr.19: 36151457-36151457 on Build GRCh38
rs564486987 Chr.19: 36151407-36151407 on Build GRCh38
rs185906339 Chr.19: 36151437-36151437 on Build GRCh38
rs201243582 Chr.19: 36151516-36151516 on Build GRCh38
rs774928409 Chr.19: 36151503-36151504 on Build GRCh38
rs369936523 Chr.19: 36151446-36151446 on Build GRCh38
rs888693089 Chr.19: 36151417-36151417 on Build GRCh38
rs2008683 Chr.19: 36151408-36151408 on Build GRCh38
rs768094654 Chr.19: 36151494-36151494 on Build GRCh38
rs1010930947 Chr.19: 36151345-36151345 on Build GRCh38
rs763986935 Chr.19: 36151549-36151549 on Build GRCh38
rs777804547 Chr.19: 36151530-36151530 on Build GRCh38
rs767432844 Chr.19: 36151416-36151416 on Build GRCh38
rs375983153 Chr.19: 36151498-36151498 on Build GRCh38

View Full Product Details