Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87605079-87605304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPK1
Entrez Gene ID
1612
Gene Name
death associated protein kinase 1
Gene Aliases
DAPK
UniGene
Hs.380277
Gene Chromosome Location
Chr.9: 87497228-87708634 on Build GRCh38
Transcripts
NM_001288729.1 NM_001288730.1 NM_001288731.1 NM_004938.3 XM_005251757.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4375615 c.227T>C p.V76A Chr.9: 87605118-87605118 on Build GRCh38
4837228 c.260C>T p.T87M Chr.9: 87605151-87605151 on Build GRCh38
5998681 c.264C>T p.D88D Chr.9: 87605155-87605155 on Build GRCh38
213008 c.265G>A p.V89I Chr.9: 87605156-87605156 on Build GRCh38
456193 c.273G>T p.L91L Chr.9: 87605164-87605164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771881600 Chr.9: 87605201-87605201 on Build GRCh38
rs774303491 Chr.9: 87605114-87605114 on Build GRCh38
rs1020710747 Chr.9: 87605250-87605250 on Build GRCh38
rs764809561 Chr.9: 87605162-87605162 on Build GRCh38
rs372830870 Chr.9: 87605173-87605173 on Build GRCh38
rs748171290 Chr.9: 87605191-87605191 on Build GRCh38
rs915915444 Chr.9: 87605231-87605231 on Build GRCh38
rs373110876 Chr.9: 87605155-87605155 on Build GRCh38
rs770099426 Chr.9: 87605113-87605113 on Build GRCh38
rs563697445 Chr.9: 87605187-87605187 on Build GRCh38
rs759393262 Chr.9: 87605115-87605115 on Build GRCh38
rs901638003 Chr.9: 87605235-87605235 on Build GRCh38
rs36207430 Chr.9: 87605238-87605238 on Build GRCh38
rs36207429 Chr.9: 87605131-87605131 on Build GRCh38
rs41283675 Chr.9: 87605219-87605219 on Build GRCh38
rs373247895 Chr.9: 87605184-87605184 on Build GRCh38
rs376105560 Chr.9: 87605161-87605161 on Build GRCh38
rs200968517 Chr.9: 87605182-87605182 on Build GRCh38
rs145223348 Chr.9: 87605199-87605199 on Build GRCh38
rs56200243 Chr.9: 87605257-87605257 on Build GRCh38
rs13288504 Chr.9: 87605265-87605265 on Build GRCh38
rs377408511 Chr.9: 87605185-87605185 on Build GRCh38
rs1039382115 Chr.9: 87605229-87605229 on Build GRCh38
rs760156331 Chr.9: 87605151-87605151 on Build GRCh38
rs770199895 Chr.9: 87605193-87605193 on Build GRCh38
rs187082016 Chr.9: 87605218-87605218 on Build GRCh38
rs370474736 Chr.9: 87605200-87605200 on Build GRCh38
rs74593219 Chr.9: 87605264-87605264 on Build GRCh38
rs549284190 Chr.9: 87605258-87605258 on Build GRCh38
rs541023394 Chr.9: 87605156-87605156 on Build GRCh38
rs1007211513 Chr.9: 87605128-87605128 on Build GRCh38

View Full Product Details