Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202036220-202036447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD7
Entrez Gene ID
8324
Gene Name
frizzled class receptor 7
Gene Aliases
FzE3
UniGene
Hs.173859
Gene Chromosome Location
Chr.2: 202034587-202038437 on Build GRCh38
Transcripts
NM_003507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143432 c.1605G>A p.L535L Chr.2: 202036252-202036252 on Build GRCh38
6192576 c.1594A>G p.I532V Chr.2: 202036241-202036241 on Build GRCh38
4624539 c.1701C>T p.S567S Chr.2: 202036348-202036348 on Build GRCh38
6198611 c.1586T>C p.V529A Chr.2: 202036233-202036233 on Build GRCh38
221671 c.1593G>A p.M531I Chr.2: 202036240-202036240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977753392 Chr.2: 202036419-202036419 on Build GRCh38
rs974234940 Chr.2: 202036367-202036367 on Build GRCh38
rs765723806 Chr.2: 202036360-202036360 on Build GRCh38
rs759636568 Chr.2: 202036416-202036416 on Build GRCh38
rs758853443 Chr.2: 202036276-202036276 on Build GRCh38
rs769424376 Chr.2: 202036288-202036288 on Build GRCh38
rs748151902 Chr.2: 202036403-202036403 on Build GRCh38
rs528874438 Chr.2: 202036281-202036281 on Build GRCh38
rs754937674 Chr.2: 202036416-202036418 on Build GRCh38
rs759567940 Chr.2: 202036354-202036354 on Build GRCh38
rs753302518 Chr.2: 202036366-202036366 on Build GRCh38
rs925202604 Chr.2: 202036424-202036424 on Build GRCh38
rs761575550 Chr.2: 202036322-202036322 on Build GRCh38
rs772937162 Chr.2: 202036406-202036406 on Build GRCh38
rs771841493 Chr.2: 202036404-202036404 on Build GRCh38
rs748503156 Chr.2: 202036275-202036275 on Build GRCh38
rs546449437 Chr.2: 202036377-202036377 on Build GRCh38
rs745552067 Chr.2: 202036282-202036282 on Build GRCh38
rs755768740 Chr.2: 202036380-202036380 on Build GRCh38
rs534588552 Chr.2: 202036407-202036407 on Build GRCh38
rs748759824 Chr.2: 202036290-202036290 on Build GRCh38
rs368139563 Chr.2: 202036263-202036263 on Build GRCh38
rs756155956 Chr.2: 202036255-202036255 on Build GRCh38
rs750086736 Chr.2: 202036378-202036378 on Build GRCh38
rs372045596 Chr.2: 202036289-202036289 on Build GRCh38
rs1015839962 Chr.2: 202036337-202036337 on Build GRCh38
rs771894898 Chr.2: 202036332-202036332 on Build GRCh38
rs199589707 Chr.2: 202036389-202036389 on Build GRCh38
rs754291190 Chr.2: 202036265-202036265 on Build GRCh38
rs1033879510 Chr.2: 202036374-202036374 on Build GRCh38
rs773419766 Chr.2: 202036336-202036336 on Build GRCh38
rs371345546 Chr.2: 202036382-202036382 on Build GRCh38
rs565714072 Chr.2: 202036340-202036340 on Build GRCh38
rs774179533 Chr.2: 202036303-202036303 on Build GRCh38
rs766526311 Chr.2: 202036342-202036342 on Build GRCh38
rs755458386 Chr.2: 202036267-202036267 on Build GRCh38
rs112052844 Chr.2: 202036422-202036422 on Build GRCh38
rs779576113 Chr.2: 202036384-202036384 on Build GRCh38
rs367875554 Chr.2: 202036410-202036410 on Build GRCh38
rs766517331 Chr.2: 202036260-202036260 on Build GRCh38
rs750694306 Chr.2: 202036246-202036246 on Build GRCh38
rs111263028 Chr.2: 202036273-202036273 on Build GRCh38
rs373957083 Chr.2: 202036386-202036386 on Build GRCh38
rs768569299 Chr.2: 202036296-202036296 on Build GRCh38
rs758800464 Chr.2: 202036375-202036375 on Build GRCh38
rs771147451 Chr.2: 202036408-202036408 on Build GRCh38

View Full Product Details