Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57274224-57274451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5M1
Entrez Gene ID
55745
Gene Name
adaptor related protein complex 5 mu 1 subunit
Gene Aliases
C14orf108, MUDENG, Mu5, MuD
UniGene
Hs.597349
Gene Chromosome Location
Chr.14: 57268888-57298742 on Build GRCh38
Transcripts
NM_018229.3 NR_026895.1 XM_006720196.2 XM_011536940.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4845957 c.91G>A p.E31K Chr.14: 57274260-57274260 on Build GRCh38
4051401 c.141A>G p.E47E Chr.14: 57274310-57274310 on Build GRCh38
6001396 c.191A>T p.D64V Chr.14: 57274360-57274360 on Build GRCh38
4051402 c.183A>G p.L61L Chr.14: 57274352-57274352 on Build GRCh38
4777538 c.101C>G p.A34G Chr.14: 57274270-57274270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761043919 Chr.14: 57274286-57274286 on Build GRCh38
rs201721178 Chr.14: 57274379-57274379 on Build GRCh38
rs771361986 Chr.14: 57274271-57274272 on Build GRCh38
rs368232524 Chr.14: 57274298-57274298 on Build GRCh38
rs781230099 Chr.14: 57274330-57274330 on Build GRCh38
rs751330068 Chr.14: 57274322-57274322 on Build GRCh38
rs756919617 Chr.14: 57274407-57274407 on Build GRCh38
rs747623726 Chr.14: 57274262-57274262 on Build GRCh38
rs779962163 Chr.14: 57274247-57274247 on Build GRCh38
rs372166195 Chr.14: 57274337-57274337 on Build GRCh38
rs142171275 Chr.14: 57274401-57274401 on Build GRCh38
rs769110140 Chr.14: 57274288-57274288 on Build GRCh38
rs769056857 Chr.14: 57274381-57274381 on Build GRCh38
rs200077344 Chr.14: 57274412-57274412 on Build GRCh38
rs114072619 Chr.14: 57274246-57274246 on Build GRCh38
rs998732930 Chr.14: 57274254-57274254 on Build GRCh38
rs763954006 Chr.14: 57274422-57274422 on Build GRCh38
rs747323641 Chr.14: 57274380-57274380 on Build GRCh38
rs770560807 Chr.14: 57274375-57274375 on Build GRCh38
rs200799961 Chr.14: 57274312-57274312 on Build GRCh38
rs752677166 Chr.14: 57274414-57274414 on Build GRCh38
rs763290804 Chr.14: 57274315-57274315 on Build GRCh38
rs752734335 Chr.14: 57274332-57274332 on Build GRCh38
rs781283085 Chr.14: 57274260-57274260 on Build GRCh38
rs202191261 Chr.14: 57274384-57274384 on Build GRCh38
rs751435806 Chr.14: 57274256-57274256 on Build GRCh38
rs759386854 Chr.14: 57274405-57274405 on Build GRCh38
rs915309358 Chr.14: 57274314-57274314 on Build GRCh38
rs201278202 Chr.14: 57274397-57274397 on Build GRCh38
rs749010126 Chr.14: 57274274-57274274 on Build GRCh38
rs766520114 Chr.14: 57274319-57274319 on Build GRCh38
rs544958915 Chr.14: 57274420-57274420 on Build GRCh38
rs772320322 Chr.14: 57274282-57274282 on Build GRCh38
rs898253263 Chr.14: 57274304-57274304 on Build GRCh38
rs754902688 Chr.14: 57274257-57274257 on Build GRCh38
rs761857810 Chr.14: 57274307-57274307 on Build GRCh38
rs765398350 Chr.14: 57274311-57274311 on Build GRCh38
rs753324122 Chr.14: 57274426-57274426 on Build GRCh38
rs148346858 Chr.14: 57274373-57274373 on Build GRCh38
rs777272858 Chr.14: 57274273-57274273 on Build GRCh38
rs766586055 Chr.14: 57274398-57274398 on Build GRCh38
rs556156925 Chr.14: 57274396-57274396 on Build GRCh38
rs777090525 Chr.14: 57274383-57274383 on Build GRCh38
rs143414348 Chr.14: 57274336-57274336 on Build GRCh38
rs775983041 Chr.14: 57274285-57274285 on Build GRCh38
rs889278862 Chr.14: 57274374-57274374 on Build GRCh38
rs748880199 Chr.14: 57274371-57274371 on Build GRCh38
rs747476401 Chr.14: 57274266-57274266 on Build GRCh38
rs374333598 Chr.14: 57274267-57274267 on Build GRCh38
rs754853892 Chr.14: 57274325-57274325 on Build GRCh38

View Full Product Details