Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 122422605-122422838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF2
Entrez Gene ID
3298
Gene Name
heat shock transcription factor 2
Gene Aliases
HSF 2, HSTF 2
UniGene
Hs.158195
Gene Chromosome Location
Chr.6: 122399551-122433119 on Build GRCh38
Transcripts
NM_001135564.1 NM_004506.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
128199 c.894C>T p.V298V Chr.6: 122422781-122422781 on Build GRCh38
4845647 c.900G>A p.Q300Q Chr.6: 122422787-122422787 on Build GRCh38
1132044 c.833G>T p.C278F Chr.6: 122422720-122422720 on Build GRCh38
1187147 c.839A>C p.Q280P Chr.6: 122422726-122422726 on Build GRCh38
5869219 c.944G>A p.G315D Chr.6: 122422831-122422831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549663651 Chr.6: 122422659-122422659 on Build GRCh38
rs145103558 Chr.6: 122422746-122422746 on Build GRCh38
rs867717077 Chr.6: 122422704-122422704 on Build GRCh38
rs774480168 Chr.6: 122422759-122422759 on Build GRCh38
rs569942295 Chr.6: 122422685-122422685 on Build GRCh38
rs765081372 Chr.6: 122422758-122422758 on Build GRCh38
rs764752770 Chr.6: 122422815-122422815 on Build GRCh38
rs910638629 Chr.6: 122422638-122422638 on Build GRCh38
rs943420074 Chr.6: 122422642-122422642 on Build GRCh38
rs888164307 Chr.6: 122422708-122422708 on Build GRCh38
rs752826332 Chr.6: 122422781-122422781 on Build GRCh38
rs570240423 Chr.6: 122422671-122422671 on Build GRCh38
rs1042675140 Chr.6: 122422730-122422730 on Build GRCh38
rs768585042 Chr.6: 122422745-122422745 on Build GRCh38
rs370934954 Chr.6: 122422689-122422689 on Build GRCh38
rs761574648 Chr.6: 122422748-122422748 on Build GRCh38
rs775005039 Chr.6: 122422726-122422726 on Build GRCh38
rs771313088 Chr.6: 122422691-122422691 on Build GRCh38
rs201784385 Chr.6: 122422775-122422775 on Build GRCh38
rs771529420 Chr.6: 122422709-122422709 on Build GRCh38
rs759785469 Chr.6: 122422763-122422763 on Build GRCh38
rs745500515 Chr.6: 122422703-122422703 on Build GRCh38
rs267600788 Chr.6: 122422731-122422731 on Build GRCh38
rs778497573 Chr.6: 122422696-122422696 on Build GRCh38
rs140579788 Chr.6: 122422790-122422790 on Build GRCh38
rs368381387 Chr.6: 122422672-122422672 on Build GRCh38
rs984731871 Chr.6: 122422631-122422632 on Build GRCh38
rs767805539 Chr.6: 122422773-122422773 on Build GRCh38
rs766865019 Chr.6: 122422724-122422724 on Build GRCh38

View Full Product Details