Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138513499-138513764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETF1
Entrez Gene ID
2107
Gene Name
eukaryotic translation termination factor 1
Gene Aliases
D5S1995, ERF, ERF1, RF1, SUP45L1, TB3-1
UniGene
Hs.483494
Gene Chromosome Location
Chr.5: 138506093-138543300 on Build GRCh38
Transcripts
NM_001256302.1 NM_001282185.1 NM_001291974.1 NM_001291975.1 NM_004730.3 XM_005271921.1 XM_017009221.1 XM_017009222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6337068 c.510C>A p.H170Q Chr.5: 138513599-138513599 on Build GRCh38
4646115 c.404C>T p.A135V Chr.5: 138513705-138513705 on Build GRCh38
3237466 c.419T>C p.L140P Chr.5: 138513690-138513690 on Build GRCh38
3609020 c.516C>T p.F172F Chr.5: 138513593-138513593 on Build GRCh38
6250297 c.541+7C>A p.? Chr.5: 138513561-138513561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967033350 Chr.5: 138513590-138513590 on Build GRCh38
rs779295866 Chr.5: 138513569-138513569 on Build GRCh38
rs912342193 Chr.5: 138513720-138513720 on Build GRCh38
rs750914121 Chr.5: 138513540-138513540 on Build GRCh38
rs199800503 Chr.5: 138513528-138513528 on Build GRCh38
rs754246870 Chr.5: 138513560-138513560 on Build GRCh38
rs754342970 Chr.5: 138513740-138513740 on Build GRCh38
rs199838059 Chr.5: 138513635-138513635 on Build GRCh38
rs201164792 Chr.5: 138513533-138513533 on Build GRCh38
rs769510937 Chr.5: 138513698-138513698 on Build GRCh38
rs770871708 Chr.5: 138513628-138513628 on Build GRCh38
rs552524795 Chr.5: 138513522-138513522 on Build GRCh38
rs756619047 Chr.5: 138513549-138513549 on Build GRCh38
rs763558454 Chr.5: 138513732-138513732 on Build GRCh38
rs761149356 Chr.5: 138513734-138513734 on Build GRCh38
rs375041421 Chr.5: 138513558-138513558 on Build GRCh38
rs746970960 Chr.5: 138513623-138513623 on Build GRCh38
rs944209433 Chr.5: 138513724-138513724 on Build GRCh38
rs779915105 Chr.5: 138513564-138513565 on Build GRCh38
rs775154168 Chr.5: 138513706-138513706 on Build GRCh38
rs376812368 Chr.5: 138513739-138513739 on Build GRCh38
rs367751940 Chr.5: 138513572-138513572 on Build GRCh38
rs534818676 Chr.5: 138513611-138513611 on Build GRCh38
rs777863501 Chr.5: 138513617-138513617 on Build GRCh38
rs762542658 Chr.5: 138513730-138513730 on Build GRCh38
rs759313013 Chr.5: 138513680-138513680 on Build GRCh38
rs372759180 Chr.5: 138513733-138513733 on Build GRCh38

View Full Product Details