Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96784711-96784984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPQ
Entrez Gene ID
10404
Gene Name
carboxypeptidase Q
Gene Aliases
LDP, PGCP
UniGene
Hs.156178
Gene Chromosome Location
Chr.8: 96645227-97143503 on Build GRCh38
Transcripts
NM_016134.3 XM_017012953.1 XM_017012954.1 XM_017012955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3942833 c.9C>G p.F3L Chr.8: 96784906-96784906 on Build GRCh38
269875 c.19G>A p.A7T Chr.8: 96784916-96784916 on Build GRCh38
1102781 c.59G>T p.G20V Chr.8: 96784956-96784956 on Build GRCh38
5951922 c.27C>T p.F9F Chr.8: 96784924-96784924 on Build GRCh38
5076257 c.86C>A p.S29Y Chr.8: 96784983-96784983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040977007 Chr.8: 96784767-96784767 on Build GRCh38
rs763907056 Chr.8: 96784882-96784882 on Build GRCh38
rs886217606 Chr.8: 96784745-96784745 on Build GRCh38
rs140523552 Chr.8: 96784736-96784736 on Build GRCh38
rs1046210621 Chr.8: 96784808-96784808 on Build GRCh38
rs893380227 Chr.8: 96784842-96784842 on Build GRCh38
rs142316711 Chr.8: 96784816-96784816 on Build GRCh38
rs916933272 Chr.8: 96784950-96784960 on Build GRCh38
rs183172032 Chr.8: 96784916-96784916 on Build GRCh38
rs1007730288 Chr.8: 96784901-96784901 on Build GRCh38
rs200887903 Chr.8: 96784894-96784894 on Build GRCh38
rs747054913 Chr.8: 96784956-96784956 on Build GRCh38
rs765027440 Chr.8: 96784899-96784899 on Build GRCh38
rs772446810 Chr.8: 96784944-96784944 on Build GRCh38
rs903283339 Chr.8: 96784831-96784831 on Build GRCh38
rs555304971 Chr.8: 96784748-96784748 on Build GRCh38
rs191258208 Chr.8: 96784900-96784900 on Build GRCh38
rs765822863 Chr.8: 96784915-96784915 on Build GRCh38
rs761498745 Chr.8: 96784898-96784898 on Build GRCh38
rs758074689 Chr.8: 96784912-96784912 on Build GRCh38
rs781070792 Chr.8: 96784890-96784890 on Build GRCh38
rs575239324 Chr.8: 96784832-96784832 on Build GRCh38
rs746048796 Chr.8: 96784939-96784939 on Build GRCh38
rs369831003 Chr.8: 96784925-96784925 on Build GRCh38
rs949250758 Chr.8: 96784775-96784775 on Build GRCh38
rs959156328 Chr.8: 96784938-96784938 on Build GRCh38
rs900644565 Chr.8: 96784824-96784824 on Build GRCh38
rs937949272 Chr.8: 96784734-96784734 on Build GRCh38
rs775821667 Chr.8: 96784951-96784951 on Build GRCh38
rs746815323 Chr.8: 96784924-96784924 on Build GRCh38
rs569818798 Chr.8: 96784825-96784825 on Build GRCh38
rs201719687 Chr.8: 96784889-96784890 on Build GRCh38
rs745946672 Chr.8: 96784919-96784919 on Build GRCh38
rs150845839 Chr.8: 96784836-96784836 on Build GRCh38
rs760611528 Chr.8: 96784879-96784879 on Build GRCh38
rs201861702 Chr.8: 96784935-96784935 on Build GRCh38

View Full Product Details