Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44794139-44794394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390079 c.1356G>A p.P452P Chr.19: 44794275-44794275 on Build GRCh38
6620918 c.1332_1333insA p.R445fs*>31 Chr.19: 44794251-44794252 on Build GRCh38
5020655 c.1303C>T p.P435S Chr.19: 44794222-44794222 on Build GRCh38
1750969 c.1324C>G p.L442V Chr.19: 44794243-44794243 on Build GRCh38
4463409 c.1290C>T p.P430P Chr.19: 44794209-44794209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770035168 Chr.19: 44794235-44794235 on Build GRCh38
rs756784889 Chr.19: 44794206-44794206 on Build GRCh38
rs764791471 Chr.19: 44794284-44794284 on Build GRCh38
rs557733635 Chr.19: 44794216-44794216 on Build GRCh38
rs753825185 Chr.19: 44794282-44794282 on Build GRCh38
rs148527231 Chr.19: 44794275-44794275 on Build GRCh38
rs959883294 Chr.19: 44794333-44794333 on Build GRCh38
rs766524066 Chr.19: 44794266-44794266 on Build GRCh38
rs377099059 Chr.19: 44794288-44794288 on Build GRCh38
rs777033769 Chr.19: 44794227-44794227 on Build GRCh38
rs757152687 Chr.19: 44794283-44794283 on Build GRCh38
rs114699167 Chr.19: 44794337-44794337 on Build GRCh38
rs773422704 Chr.19: 44794250-44794250 on Build GRCh38
rs374088584 Chr.19: 44794199-44794200 on Build GRCh38
rs1903831 Chr.19: 44794197-44794197 on Build GRCh38
rs369770573 Chr.19: 44794311-44794311 on Build GRCh38
rs752421980 Chr.19: 44794198-44794198 on Build GRCh38
rs371869408 Chr.19: 44794197-44794197 on Build GRCh38
rs748733485 Chr.19: 44794317-44794317 on Build GRCh38
rs763325522 Chr.19: 44794259-44794259 on Build GRCh38
rs779576249 Chr.19: 44794293-44794293 on Build GRCh38
rs758039759 Chr.19: 44794215-44794215 on Build GRCh38
rs34259391 Chr.19: 44794193-44794194 on Build GRCh38
rs200260767 Chr.19: 44794194-44794194 on Build GRCh38
rs1041110421 Chr.19: 44794192-44794192 on Build GRCh38
rs138476890 Chr.19: 44794268-44794268 on Build GRCh38
rs777137191 Chr.19: 44794162-44794162 on Build GRCh38
rs757863605 Chr.19: 44794292-44794292 on Build GRCh38
rs781596263 Chr.19: 44794314-44794314 on Build GRCh38
rs375340184 Chr.19: 44794177-44794177 on Build GRCh38
rs1020506607 Chr.19: 44794355-44794355 on Build GRCh38
rs761988158 Chr.19: 44794163-44794163 on Build GRCh38
rs537580232 Chr.19: 44794201-44794201 on Build GRCh38
rs374048625 Chr.19: 44794234-44794234 on Build GRCh38
rs386809696 Chr.19: 44794197-44794199 on Build GRCh38
rs77416350 Chr.19: 44794199-44794199 on Build GRCh38
rs778376496 Chr.19: 44794209-44794209 on Build GRCh38
rs369262197 Chr.19: 44794200-44794200 on Build GRCh38
rs997076042 Chr.19: 44794271-44794271 on Build GRCh38
rs768998253 Chr.19: 44794224-44794224 on Build GRCh38
rs1010185585 Chr.19: 44794298-44794298 on Build GRCh38
rs745556828 Chr.19: 44794214-44794214 on Build GRCh38
rs775349494 Chr.19: 44794304-44794304 on Build GRCh38
rs762117393 Chr.19: 44794181-44794183 on Build GRCh38
rs374667364 Chr.19: 44794322-44794322 on Build GRCh38
rs766555920 Chr.19: 44794245-44794246 on Build GRCh38
rs116023028 Chr.19: 44794222-44794222 on Build GRCh38

View Full Product Details