Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31687863-31688106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD16A
Entrez Gene ID
7920
Gene Name
abhydrolase domain containing 16A
Gene Aliases
BAT5, D6S82E, NG26, PP199
UniGene
Hs.388188
Gene Chromosome Location
Chr.6: 31686949-31703360 on Build GRCh38
Transcripts
NM_001177515.1 NM_021160.2 NR_033488.1 NR_033489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5767270 c.1390G>C p.E464Q Chr.6: 31687881-31687881 on Build GRCh38
3346388 c.1381G>C p.V461L Chr.6: 31687890-31687890 on Build GRCh38
7115988 c.1312C>G p.P438A Chr.6: 31688099-31688099 on Build GRCh38
4354153 c.1402C>T p.R468* Chr.6: 31687869-31687869 on Build GRCh38
5011602 c.1375C>T p.P459S Chr.6: 31687896-31687896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763973759 Chr.6: 31687936-31687936 on Build GRCh38
rs767867426 Chr.6: 31687934-31687935 on Build GRCh38
rs760211117 Chr.6: 31688067-31688067 on Build GRCh38
rs745796053 Chr.6: 31687916-31687916 on Build GRCh38
rs371203333 Chr.6: 31687905-31687905 on Build GRCh38
rs775502904 Chr.6: 31687935-31687935 on Build GRCh38
rs749286376 Chr.6: 31688030-31688030 on Build GRCh38
rs116996610 Chr.6: 31687904-31687904 on Build GRCh38
rs955194965 Chr.6: 31688071-31688071 on Build GRCh38
rs201502942 Chr.6: 31687890-31687890 on Build GRCh38
rs774259910 Chr.6: 31688041-31688041 on Build GRCh38
rs1019677730 Chr.6: 31687956-31687956 on Build GRCh38
rs879234031 Chr.6: 31687971-31687971 on Build GRCh38
rs747025358 Chr.6: 31688022-31688022 on Build GRCh38
rs549698751 Chr.6: 31687995-31687995 on Build GRCh38
rs781565726 Chr.6: 31688020-31688020 on Build GRCh38
rs981164124 Chr.6: 31688066-31688066 on Build GRCh38
rs773025757 Chr.6: 31688064-31688064 on Build GRCh38
rs34796493 Chr.6: 31687892-31687893 on Build GRCh38
rs768330442 Chr.6: 31688034-31688034 on Build GRCh38
rs531666335 Chr.6: 31687909-31687909 on Build GRCh38
rs773842168 Chr.6: 31687938-31687938 on Build GRCh38
rs571465940 Chr.6: 31688018-31688018 on Build GRCh38
rs138712884 Chr.6: 31688019-31688019 on Build GRCh38
rs754270788 Chr.6: 31688079-31688079 on Build GRCh38
rs777983993 Chr.6: 31688021-31688021 on Build GRCh38
rs755686569 Chr.6: 31688002-31688002 on Build GRCh38
rs1000470101 Chr.6: 31688011-31688011 on Build GRCh38
rs775243440 Chr.6: 31688025-31688025 on Build GRCh38
rs547919675 Chr.6: 31688077-31688077 on Build GRCh38
rs376190924 Chr.6: 31688024-31688024 on Build GRCh38
rs906088328 Chr.6: 31687987-31687987 on Build GRCh38
rs761624021 Chr.6: 31687990-31687990 on Build GRCh38
rs769661318 Chr.6: 31687926-31687926 on Build GRCh38
rs74356397 Chr.6: 31687892-31687892 on Build GRCh38
rs910892425 Chr.6: 31688082-31688082 on Build GRCh38
rs761713972 Chr.6: 31688042-31688042 on Build GRCh38
rs994166609 Chr.6: 31688008-31688008 on Build GRCh38
rs766752943 Chr.6: 31688014-31688014 on Build GRCh38
rs767208638 Chr.6: 31688055-31688055 on Build GRCh38
rs778951516 Chr.6: 31687893-31687893 on Build GRCh38

View Full Product Details