Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17119615-17119860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR6
Entrez Gene ID
27439
Gene Name
cat eye syndrome chromosome region, candidate 6
Gene Aliases
-
UniGene
Hs.209577
Gene Chromosome Location
Chr.22: 17116299-17121367 on Build GRCh38
Transcripts
NM_001163079.1 NM_031890.3 XM_011546124.2 XM_011546125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7265646 c.1510C>A p.L504I Chr.22: 17119618-17119618 on Build GRCh38
4648509 c.1447G>A p.A483T Chr.22: 17119681-17119681 on Build GRCh38
5033338 c.1369G>A p.G457S Chr.22: 17119759-17119759 on Build GRCh38
5761442 c.1343C>T p.A448V Chr.22: 17119785-17119785 on Build GRCh38
369085 c.1341C>T p.N447N Chr.22: 17119787-17119787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867203839 Chr.22: 17119786-17119786 on Build GRCh38
rs939826374 Chr.22: 17119823-17119823 on Build GRCh38
rs371550781 Chr.22: 17119793-17119793 on Build GRCh38
rs778427237 Chr.22: 17119824-17119824 on Build GRCh38
rs777950085 Chr.22: 17119719-17119719 on Build GRCh38
rs61741012 Chr.22: 17119799-17119799 on Build GRCh38
rs768546783 Chr.22: 17119748-17119748 on Build GRCh38
rs759784023 Chr.22: 17119796-17119796 on Build GRCh38
rs760185615 Chr.22: 17119792-17119792 on Build GRCh38
rs765592655 Chr.22: 17119797-17119797 on Build GRCh38
rs755425657 Chr.22: 17119676-17119676 on Build GRCh38
rs368814541 Chr.22: 17119838-17119838 on Build GRCh38
rs61741055 Chr.22: 17119782-17119782 on Build GRCh38
rs761770456 Chr.22: 17119777-17119777 on Build GRCh38
rs754138784 Chr.22: 17119653-17119653 on Build GRCh38
rs749025270 Chr.22: 17119746-17119746 on Build GRCh38
rs781627390 Chr.22: 17119744-17119744 on Build GRCh38
rs753283216 Chr.22: 17119699-17119699 on Build GRCh38
rs12628507 Chr.22: 17119809-17119809 on Build GRCh38
rs778992648 Chr.22: 17119820-17119820 on Build GRCh38
rs757471641 Chr.22: 17119834-17119834 on Build GRCh38
rs747098498 Chr.22: 17119722-17119722 on Build GRCh38
rs999242123 Chr.22: 17119737-17119737 on Build GRCh38
rs764730634 Chr.22: 17119693-17119693 on Build GRCh38
rs771209447 Chr.22: 17119736-17119736 on Build GRCh38
rs970648258 Chr.22: 17119787-17119787 on Build GRCh38
rs758856625 Chr.22: 17119718-17119718 on Build GRCh38
rs564252715 Chr.22: 17119784-17119784 on Build GRCh38
rs774126006 Chr.22: 17119773-17119773 on Build GRCh38
rs758882475 Chr.22: 17119811-17119811 on Build GRCh38
rs779263034 Chr.22: 17119695-17119695 on Build GRCh38
rs867445414 Chr.22: 17119683-17119683 on Build GRCh38
rs572237782 Chr.22: 17119685-17119685 on Build GRCh38
rs543023653 Chr.22: 17119829-17119829 on Build GRCh38
rs553697504 Chr.22: 17119645-17119645 on Build GRCh38

View Full Product Details