Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56349723-56349960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASDH
Entrez Gene ID
132949
Gene Name
aminoadipate-semialdehyde dehydrogenase
Gene Aliases
ACSF4, LYS2, NRPS1098, NRPS998
UniGene
Hs.104347
Gene Chromosome Location
Chr.4: 56337687-56387677 on Build GRCh38
Transcripts
NM_001286668.1 NM_001286669.1 NM_001286670.1 NM_001286671.1 NM_001286672.1 NM_001323890.1 NM_001323892.1 NM_001323893.1 NM_001323899.1 NM_181806.3 XM_017007740.1 XM_017007741.1 XM_017007742.1 XM_017007743.1 XM_017007744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4382268 c.1811G>A p.G604D Chr.4: 56349940-56349940 on Build GRCh38
1430296 c.1797T>C p.I599I Chr.4: 56349954-56349954 on Build GRCh38
6856114 c.1915A>G p.R639G Chr.4: 56349836-56349836 on Build GRCh38
4871961 c.1816T>A p.S606T Chr.4: 56349935-56349935 on Build GRCh38
3604645 c.1957C>T p.Q653* Chr.4: 56349794-56349794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764856065 Chr.4: 56349767-56349767 on Build GRCh38
rs571428751 Chr.4: 56349899-56349899 on Build GRCh38
rs747374372 Chr.4: 56349873-56349873 on Build GRCh38
rs369811234 Chr.4: 56349896-56349896 on Build GRCh38
rs769149976 Chr.4: 56349901-56349901 on Build GRCh38
rs534415378 Chr.4: 56349752-56349752 on Build GRCh38
rs774959231 Chr.4: 56349782-56349782 on Build GRCh38
rs746308276 Chr.4: 56349781-56349781 on Build GRCh38
rs138653291 Chr.4: 56349860-56349860 on Build GRCh38
rs151197679 Chr.4: 56349824-56349824 on Build GRCh38
rs758708471 Chr.4: 56349864-56349864 on Build GRCh38
rs891017676 Chr.4: 56349875-56349875 on Build GRCh38
rs754991127 Chr.4: 56349836-56349836 on Build GRCh38
rs541811269 Chr.4: 56349885-56349885 on Build GRCh38
rs773718786 Chr.4: 56349755-56349755 on Build GRCh38
rs772779965 Chr.4: 56349928-56349928 on Build GRCh38
rs774053165 Chr.4: 56349930-56349930 on Build GRCh38
rs932622420 Chr.4: 56349831-56349831 on Build GRCh38
rs758508593 Chr.4: 56349824-56349825 on Build GRCh38
rs758941902 Chr.4: 56349763-56349763 on Build GRCh38
rs183549178 Chr.4: 56349868-56349868 on Build GRCh38
rs61978622 Chr.4: 56349803-56349803 on Build GRCh38
rs769442659 Chr.4: 56349867-56349867 on Build GRCh38
rs144223835 Chr.4: 56349756-56349756 on Build GRCh38
rs762453278 Chr.4: 56349907-56349907 on Build GRCh38
rs775067055 Chr.4: 56349902-56349902 on Build GRCh38
rs779070617 Chr.4: 56349846-56349846 on Build GRCh38
rs377506396 Chr.4: 56349883-56349883 on Build GRCh38
rs776986545 Chr.4: 56349888-56349888 on Build GRCh38
rs776659864 Chr.4: 56349764-56349764 on Build GRCh38
rs763808403 Chr.4: 56349804-56349804 on Build GRCh38
rs763719524 Chr.4: 56349915-56349915 on Build GRCh38
rs751367190 Chr.4: 56349808-56349808 on Build GRCh38
rs143160554 Chr.4: 56349823-56349823 on Build GRCh38
rs777345008 Chr.4: 56349758-56349758 on Build GRCh38

View Full Product Details