Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 148651854-148652105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF2
Entrez Gene ID
2334
Gene Name
AF4/FMR2 family member 2
Gene Aliases
FMR2, FMR2P, FRAXE, MRX2, OX19
UniGene
Hs.496911
Gene Chromosome Location
Chr.X: 148500619-149000663 on Build GRCh38
Transcripts
NM_001169122.1 NM_001169123.1 NM_001169124.1 NM_001169125.1 NM_002025.3

SNP Detail

SNP ID Chromosome Location
rs782455801 Chr.X: 148651950-148651950 on Build GRCh38
rs372514798 Chr.X: 148652015-148652015 on Build GRCh38
rs782431970 Chr.X: 148652035-148652035 on Build GRCh38
rs782049601 Chr.X: 148652016-148652016 on Build GRCh38
rs909283684 Chr.X: 148652018-148652018 on Build GRCh38
rs144497911 Chr.X: 148652056-148652056 on Build GRCh38
rs782625275 Chr.X: 148651994-148651994 on Build GRCh38
rs781827532 Chr.X: 148652059-148652059 on Build GRCh38
rs782340089 Chr.X: 148652014-148652014 on Build GRCh38
rs782225647 Chr.X: 148652009-148652009 on Build GRCh38
rs143135907 Chr.X: 148652045-148652045 on Build GRCh38
rs41312622 Chr.X: 148651990-148651990 on Build GRCh38
rs139180816 Chr.X: 148651968-148651968 on Build GRCh38
rs782129403 Chr.X: 148652046-148652046 on Build GRCh38
rs782561675 Chr.X: 148652074-148652074 on Build GRCh38
rs782680011 Chr.X: 148651960-148651960 on Build GRCh38

View Full Product Details