Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32424480-32424753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO1
Entrez Gene ID
48
Gene Name
aconitase 1
Gene Aliases
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1
UniGene
Hs.567229
Gene Chromosome Location
Chr.9: 32384603-32450836 on Build GRCh38
Transcripts
NM_001278352.1 NM_002197.2 XM_005251476.1 XM_011517888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1674445 c.1132G>A p.D378N Chr.9: 32424609-32424609 on Build GRCh38
2771968 c.1117C>T p.P373S Chr.9: 32424594-32424594 on Build GRCh38
6592236 c.1133A>G p.D378G Chr.9: 32424610-32424610 on Build GRCh38
6592215 c.1165T>G p.F389V Chr.9: 32424642-32424642 on Build GRCh38
6592207 c.1149C>T p.S383S Chr.9: 32424626-32424626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778340187 Chr.9: 32424620-32424620 on Build GRCh38
rs780944307 Chr.9: 32424665-32424665 on Build GRCh38
rs772943365 Chr.9: 32424688-32424688 on Build GRCh38
rs919301179 Chr.9: 32424534-32424534 on Build GRCh38
rs775380821 Chr.9: 32424522-32424522 on Build GRCh38
rs571429620 Chr.9: 32424678-32424678 on Build GRCh38
rs749518811 Chr.9: 32424672-32424672 on Build GRCh38
rs201983073 Chr.9: 32424550-32424550 on Build GRCh38
rs781637839 Chr.9: 32424650-32424650 on Build GRCh38
rs750063171 Chr.9: 32424675-32424675 on Build GRCh38
rs768655335 Chr.9: 32424538-32424538 on Build GRCh38
rs757907809 Chr.9: 32424635-32424635 on Build GRCh38
rs752097488 Chr.9: 32424626-32424626 on Build GRCh38
rs3814519 Chr.9: 32424661-32424661 on Build GRCh38
rs114405511 Chr.9: 32424726-32424726 on Build GRCh38
rs770099296 Chr.9: 32424667-32424667 on Build GRCh38
rs764802765 Chr.9: 32424711-32424711 on Build GRCh38
rs780288187 Chr.9: 32424669-32424669 on Build GRCh38
rs1026989862 Chr.9: 32424513-32424513 on Build GRCh38
rs200462190 Chr.9: 32424597-32424597 on Build GRCh38
rs774323213 Chr.9: 32424549-32424549 on Build GRCh38
rs7025336 Chr.9: 32424647-32424647 on Build GRCh38
rs796792969 Chr.9: 32424529-32424529 on Build GRCh38
rs377094927 Chr.9: 32424698-32424698 on Build GRCh38
rs754480816 Chr.9: 32424619-32424619 on Build GRCh38
rs772009189 Chr.9: 32424686-32424686 on Build GRCh38
rs200236829 Chr.9: 32424533-32424533 on Build GRCh38
rs570643968 Chr.9: 32424598-32424598 on Build GRCh38
rs767294022 Chr.9: 32424563-32424563 on Build GRCh38
rs947775422 Chr.9: 32424687-32424687 on Build GRCh38
rs746087542 Chr.9: 32424506-32424506 on Build GRCh38
rs750222926 Chr.9: 32424585-32424585 on Build GRCh38
rs376937935 Chr.9: 32424703-32424703 on Build GRCh38
rs889604794 Chr.9: 32424708-32424708 on Build GRCh38
rs924145609 Chr.9: 32424627-32424627 on Build GRCh38
rs960121496 Chr.9: 32424594-32424594 on Build GRCh38
rs760265120 Chr.9: 32424586-32424586 on Build GRCh38
rs965681864 Chr.9: 32424612-32424612 on Build GRCh38
rs766080386 Chr.9: 32424590-32424590 on Build GRCh38
rs549050192 Chr.9: 32424512-32424512 on Build GRCh38
rs1005396696 Chr.9: 32424723-32424723 on Build GRCh38
rs761796391 Chr.9: 32424560-32424560 on Build GRCh38
rs899008256 Chr.9: 32424732-32424732 on Build GRCh38
rs368953124 Chr.9: 32424705-32424705 on Build GRCh38
rs45450994 Chr.9: 32424677-32424677 on Build GRCh38
rs1006882737 Chr.9: 32424572-32424572 on Build GRCh38
rs77785385 Chr.9: 32424700-32424700 on Build GRCh38

View Full Product Details