Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109667904-109668153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL4
Entrez Gene ID
2182
Gene Name
acyl-CoA synthetase long-chain family member 4
Gene Aliases
ACS4, FACL4, LACS4, MRX63, MRX68
UniGene
Hs.268785
Gene Chromosome Location
Chr.X: 109641330-109733403 on Build GRCh38
Transcripts
NM_001318509.1 NM_001318510.1 NM_004458.2 NM_022977.2 XM_005262109.2 XM_006724635.2 XM_011530888.2 XM_011530889.2

SNP Detail

SNP ID Chromosome Location
rs778712018 Chr.X: 109668101-109668101 on Build GRCh38
rs959630027 Chr.X: 109667943-109667943 on Build GRCh38
rs983596601 Chr.X: 109668042-109668042 on Build GRCh38
rs754719842 Chr.X: 109668062-109668062 on Build GRCh38
rs186413776 Chr.X: 109667963-109667963 on Build GRCh38
rs746094072 Chr.X: 109668054-109668054 on Build GRCh38
rs990913345 Chr.X: 109667986-109667986 on Build GRCh38
rs964976738 Chr.X: 109667975-109667975 on Build GRCh38
rs779262168 Chr.X: 109668002-109668002 on Build GRCh38
rs191072898 Chr.X: 109667976-109667976 on Build GRCh38
rs974725218 Chr.X: 109667989-109667989 on Build GRCh38
rs954571523 Chr.X: 109668005-109668005 on Build GRCh38
rs920762563 Chr.X: 109668001-109668001 on Build GRCh38
rs372786751 Chr.X: 109668118-109668118 on Build GRCh38
rs115092395 Chr.X: 109668030-109668030 on Build GRCh38
rs112736666 Chr.X: 109668117-109668117 on Build GRCh38
rs181072568 Chr.X: 109667956-109667956 on Build GRCh38
rs865808317 Chr.X: 109668112-109668112 on Build GRCh38

View Full Product Details