Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20974613-20974866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
UniGene
Hs.723246
Gene Chromosome Location
Chr.22: 20965130-20981360 on Build GRCh38
Transcripts
NM_001018060.2 NM_001146288.1 NM_144704.2 NR_027464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4557307 c.668G>A p.R223H Chr.22: 20974800-20974800 on Build GRCh38
117262 c.650G>T p.G217V Chr.22: 20974746-20974746 on Build GRCh38
4557306 c.722G>A p.R241H Chr.22: 20974800-20974800 on Build GRCh38
6597188 c.705G>A p.R235R Chr.22: 20974783-20974783 on Build GRCh38
1182312 c.677C>T p.T226M Chr.22: 20974773-20974773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199990563 Chr.22: 20974687-20974687 on Build GRCh38
rs755589476 Chr.22: 20974782-20974782 on Build GRCh38
rs768996285 Chr.22: 20974737-20974737 on Build GRCh38
rs767165531 Chr.22: 20974844-20974844 on Build GRCh38
rs751281437 Chr.22: 20974691-20974691 on Build GRCh38
rs770269459 Chr.22: 20974821-20974821 on Build GRCh38
rs762752706 Chr.22: 20974688-20974688 on Build GRCh38
rs140711648 Chr.22: 20974781-20974781 on Build GRCh38
rs759440982 Chr.22: 20974677-20974677 on Build GRCh38
rs760759626 Chr.22: 20974758-20974758 on Build GRCh38
rs373816290 Chr.22: 20974774-20974774 on Build GRCh38
rs538743781 Chr.22: 20974636-20974636 on Build GRCh38
rs769632815 Chr.22: 20974759-20974759 on Build GRCh38
rs138399322 Chr.22: 20974795-20974795 on Build GRCh38
rs772400232 Chr.22: 20974753-20974753 on Build GRCh38
rs780732378 Chr.22: 20974693-20974693 on Build GRCh38
rs766798038 Chr.22: 20974783-20974783 on Build GRCh38
rs775433268 Chr.22: 20974762-20974762 on Build GRCh38
rs756893161 Chr.22: 20974692-20974692 on Build GRCh38
rs553803201 Chr.22: 20974799-20974799 on Build GRCh38
rs779413799 Chr.22: 20974743-20974743 on Build GRCh38
rs376761164 Chr.22: 20974657-20974657 on Build GRCh38
rs77667836 Chr.22: 20974833-20974833 on Build GRCh38
rs140062749 Chr.22: 20974724-20974724 on Build GRCh38
rs558665585 Chr.22: 20974669-20974669 on Build GRCh38
rs767120379 Chr.22: 20974776-20974776 on Build GRCh38
rs758982243 Chr.22: 20974735-20974735 on Build GRCh38
rs145844241 Chr.22: 20974746-20974746 on Build GRCh38
rs178266 Chr.22: 20974714-20974714 on Build GRCh38
rs375039875 Chr.22: 20974817-20974817 on Build GRCh38
rs777993611 Chr.22: 20974800-20974800 on Build GRCh38
rs150271585 Chr.22: 20974736-20974736 on Build GRCh38
rs370089581 Chr.22: 20974757-20974757 on Build GRCh38
rs763742407 Chr.22: 20974690-20974690 on Build GRCh38
rs201527012 Chr.22: 20974802-20974802 on Build GRCh38
rs761497799 Chr.22: 20974775-20974775 on Build GRCh38
rs773410446 Chr.22: 20974676-20974676 on Build GRCh38
rs733455 Chr.22: 20974834-20974834 on Build GRCh38
rs771110770 Chr.22: 20974808-20974808 on Build GRCh38
rs202122018 Chr.22: 20974763-20974763 on Build GRCh38
rs545876573 Chr.22: 20974831-20974831 on Build GRCh38
rs373611938 Chr.22: 20974649-20974649 on Build GRCh38
rs779390732 Chr.22: 20974710-20974710 on Build GRCh38
rs373499169 Chr.22: 20974682-20974682 on Build GRCh38
rs763938239 Chr.22: 20974773-20974773 on Build GRCh38
rs761551078 Chr.22: 20974839-20974839 on Build GRCh38
rs748492260 Chr.22: 20974796-20974796 on Build GRCh38
rs1021391494 Chr.22: 20974648-20974648 on Build GRCh38
rs575044944 Chr.22: 20974794-20974794 on Build GRCh38
rs756615074 Chr.22: 20974722-20974722 on Build GRCh38
rs749298905 Chr.22: 20974822-20974822 on Build GRCh38
rs749621144 Chr.22: 20974633-20974633 on Build GRCh38
rs145844603 Chr.22: 20974792-20974792 on Build GRCh38

View Full Product Details