Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68787697-68787968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32A
Entrez Gene ID
8125
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32
UniGene
Hs.458747
Gene Chromosome Location
Chr.15: 68778535-68820922 on Build GRCh38
Transcripts
NM_006305.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6021356 c.118G>A p.E40K Chr.15: 68787856-68787856 on Build GRCh38
4756951 c.106G>A p.G36S Chr.15: 68787868-68787868 on Build GRCh38
6142881 c.172G>A p.A58T Chr.15: 68787802-68787802 on Build GRCh38
5961570 c.169A>G p.I57V Chr.15: 68787805-68787805 on Build GRCh38
4056509 c.76A>C p.N26H Chr.15: 68787898-68787898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377082441 Chr.15: 68787743-68787743 on Build GRCh38
rs769349787 Chr.15: 68787934-68787934 on Build GRCh38
rs778857524 Chr.15: 68787849-68787849 on Build GRCh38
rs778006443 Chr.15: 68787828-68787828 on Build GRCh38
rs988741555 Chr.15: 68787914-68787914 on Build GRCh38
rs774366260 Chr.15: 68787830-68787830 on Build GRCh38
rs776311932 Chr.15: 68787802-68787802 on Build GRCh38
rs761366057 Chr.15: 68787933-68787933 on Build GRCh38
rs768549533 Chr.15: 68787887-68787887 on Build GRCh38
rs772536220 Chr.15: 68787937-68787937 on Build GRCh38
rs200174860 Chr.15: 68787822-68787822 on Build GRCh38
rs745782264 Chr.15: 68787764-68787764 on Build GRCh38
rs753342674 Chr.15: 68787730-68787730 on Build GRCh38
rs956184651 Chr.15: 68787888-68787888 on Build GRCh38
rs766949764 Chr.15: 68787735-68787735 on Build GRCh38
rs370366169 Chr.15: 68787803-68787803 on Build GRCh38
rs985666071 Chr.15: 68787939-68787939 on Build GRCh38
rs755371580 Chr.15: 68787809-68787809 on Build GRCh38
rs373049653 Chr.15: 68787821-68787821 on Build GRCh38
rs763791718 Chr.15: 68787801-68787801 on Build GRCh38
rs576930598 Chr.15: 68787866-68787866 on Build GRCh38
rs775360610 Chr.15: 68787780-68787780 on Build GRCh38
rs374654173 Chr.15: 68787938-68787938 on Build GRCh38
rs754128831 Chr.15: 68787949-68787949 on Build GRCh38
rs757419555 Chr.15: 68787844-68787844 on Build GRCh38
rs199683923 Chr.15: 68787736-68787736 on Build GRCh38
rs770889423 Chr.15: 68787751-68787751 on Build GRCh38
rs373512062 Chr.15: 68787755-68787755 on Build GRCh38
rs767935426 Chr.15: 68787810-68787810 on Build GRCh38
rs753007661 Chr.15: 68787820-68787820 on Build GRCh38
rs771971476 Chr.15: 68787765-68787765 on Build GRCh38
rs186434796 Chr.15: 68787946-68787946 on Build GRCh38
rs749322303 Chr.15: 68787744-68787744 on Build GRCh38
rs748372593 Chr.15: 68787742-68787742 on Build GRCh38
rs201740562 Chr.15: 68787872-68787872 on Build GRCh38
rs368466597 Chr.15: 68787945-68787945 on Build GRCh38
rs781404728 Chr.15: 68787739-68787739 on Build GRCh38
rs181780074 Chr.15: 68787929-68787929 on Build GRCh38
rs753167154 Chr.15: 68787942-68787942 on Build GRCh38
rs756356254 Chr.15: 68787827-68787827 on Build GRCh38
rs746761102 Chr.15: 68787876-68787876 on Build GRCh38
rs752173392 Chr.15: 68787805-68787805 on Build GRCh38
rs534500785 Chr.15: 68787733-68787733 on Build GRCh38
rs760417859 Chr.15: 68787785-68787785 on Build GRCh38

View Full Product Details