Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42726002-42726257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731782 c.3247G>T p.V1083L Chr.15: 42726118-42726118 on Build GRCh38
4054740 c.3210G>T p.L1070L Chr.15: 42726155-42726155 on Build GRCh38
6988624 c.3223G>A p.E1075K Chr.15: 42726142-42726142 on Build GRCh38
7427402 c.3267C>T p.L1089L Chr.15: 42726098-42726098 on Build GRCh38
4166682 c.3214C>A p.P1072T Chr.15: 42726151-42726151 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373329905 Chr.15: 42726167-42726167 on Build GRCh38
rs181942118 Chr.15: 42726217-42726217 on Build GRCh38
rs760831544 Chr.15: 42726163-42726163 on Build GRCh38
rs753169494 Chr.15: 42726093-42726093 on Build GRCh38
rs371567068 Chr.15: 42726104-42726104 on Build GRCh38
rs371724263 Chr.15: 42726210-42726210 on Build GRCh38
rs758423857 Chr.15: 42726211-42726211 on Build GRCh38
rs923285215 Chr.15: 42726035-42726035 on Build GRCh38
rs750362336 Chr.15: 42726052-42726052 on Build GRCh38
rs993247706 Chr.15: 42726182-42726182 on Build GRCh38
rs762992673 Chr.15: 42726171-42726171 on Build GRCh38
rs575678260 Chr.15: 42726153-42726153 on Build GRCh38
rs754699165 Chr.15: 42726079-42726079 on Build GRCh38
rs778218831 Chr.15: 42726098-42726098 on Build GRCh38
rs199584270 Chr.15: 42726174-42726174 on Build GRCh38
rs375739663 Chr.15: 42726056-42726056 on Build GRCh38
rs545423135 Chr.15: 42726028-42726028 on Build GRCh38
rs370604256 Chr.15: 42726205-42726205 on Build GRCh38
rs778306555 Chr.15: 42726201-42726201 on Build GRCh38
rs758062781 Chr.15: 42726172-42726173 on Build GRCh38
rs779888204 Chr.15: 42726173-42726174 on Build GRCh38
rs769605831 Chr.15: 42726164-42726164 on Build GRCh38
rs1010288682 Chr.15: 42726082-42726082 on Build GRCh38
rs530246239 Chr.15: 42726218-42726218 on Build GRCh38
rs757822823 Chr.15: 42726202-42726202 on Build GRCh38
rs933211406 Chr.15: 42726073-42726073 on Build GRCh38
rs772939236 Chr.15: 42726165-42726165 on Build GRCh38
rs776881666 Chr.15: 42726148-42726148 on Build GRCh38
rs765932895 Chr.15: 42726061-42726061 on Build GRCh38
rs201911736 Chr.15: 42726193-42726193 on Build GRCh38
rs767955137 Chr.15: 42726196-42726197 on Build GRCh38
rs561641913 Chr.15: 42726212-42726212 on Build GRCh38
rs996376012 Chr.15: 42726074-42726074 on Build GRCh38
rs779099184 Chr.15: 42726108-42726108 on Build GRCh38
rs900718057 Chr.15: 42726040-42726040 on Build GRCh38
rs1037783864 Chr.15: 42726204-42726204 on Build GRCh38
rs745908891 Chr.15: 42726125-42726125 on Build GRCh38
rs752400883 Chr.15: 42726190-42726190 on Build GRCh38
rs749845511 Chr.15: 42726103-42726103 on Build GRCh38
rs184244219 Chr.15: 42726236-42726236 on Build GRCh38
rs772408802 Chr.15: 42726138-42726138 on Build GRCh38
rs376238524 Chr.15: 42726184-42726184 on Build GRCh38
rs780702942 Chr.15: 42726081-42726081 on Build GRCh38
rs559053820 Chr.15: 42726097-42726097 on Build GRCh38
rs1035043369 Chr.15: 42726223-42726223 on Build GRCh38
rs751007896 Chr.15: 42726075-42726075 on Build GRCh38

View Full Product Details