Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11576512-11576761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP5
Entrez Gene ID
54
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
UniGene
Hs.1211
Gene Chromosome Location
Chr.19: 11574660-11579008 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Gene Symbol
ZNF627
Entrez Gene ID
199692
Gene Name
zinc finger protein 627
Gene Aliases
-
UniGene
Hs.526665
Gene Chromosome Location
Chr.19: 11575255-11619159 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5509219 c.413G>A p.R138H Chr.19: 11576565-11576565 on Build GRCh38
4811356 c.353C>T p.S118F Chr.19: 11576752-11576752 on Build GRCh38
3796611 c.353C>T p.S118F Chr.19: 11576752-11576752 on Build GRCh38
5509218 c.413G>A p.R138H Chr.19: 11576565-11576565 on Build GRCh38
6988411 c.386G>A p.R129H Chr.19: 11576719-11576719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760906463 Chr.19: 11576601-11576601 on Build GRCh38
rs200679613 Chr.19: 11576628-11576628 on Build GRCh38
rs370365286 Chr.19: 11576678-11576678 on Build GRCh38
rs186246910 Chr.19: 11576627-11576627 on Build GRCh38
rs113937776 Chr.19: 11576652-11576652 on Build GRCh38
rs562918956 Chr.19: 11576701-11576701 on Build GRCh38
rs765472713 Chr.19: 11576613-11576613 on Build GRCh38
rs780704583 Chr.19: 11576634-11576634 on Build GRCh38
rs753781424 Chr.19: 11576643-11576643 on Build GRCh38
rs555490099 Chr.19: 11576699-11576699 on Build GRCh38
rs761983850 Chr.19: 11576609-11576609 on Build GRCh38
rs776392911 Chr.19: 11576544-11576544 on Build GRCh38
rs915517618 Chr.19: 11576546-11576546 on Build GRCh38
rs746331116 Chr.19: 11576690-11576690 on Build GRCh38
rs747619825 Chr.19: 11576736-11576736 on Build GRCh38
rs541543818 Chr.19: 11576689-11576689 on Build GRCh38
rs754840152 Chr.19: 11576631-11576631 on Build GRCh38
rs764203884 Chr.19: 11576604-11576604 on Build GRCh38
rs767853623 Chr.19: 11576585-11576585 on Build GRCh38
rs768647004 Chr.19: 11576678-11576679 on Build GRCh38
rs2305799 Chr.19: 11576536-11576536 on Build GRCh38
rs925299596 Chr.19: 11576624-11576624 on Build GRCh38
rs769232802 Chr.19: 11576640-11576640 on Build GRCh38
rs774823139 Chr.19: 11576566-11576566 on Build GRCh38
rs760044774 Chr.19: 11576568-11576568 on Build GRCh38
rs143362935 Chr.19: 11576565-11576565 on Build GRCh38
rs2328915 Chr.19: 11576647-11576647 on Build GRCh38
rs754135147 Chr.19: 11576605-11576605 on Build GRCh38
rs774986467 Chr.19: 11576705-11576705 on Build GRCh38
rs769135184 Chr.19: 11576707-11576707 on Build GRCh38
rs747855202 Chr.19: 11576636-11576636 on Build GRCh38
rs776981075 Chr.19: 11576713-11576713 on Build GRCh38
rs899103554 Chr.19: 11576700-11576700 on Build GRCh38
rs750563102 Chr.19: 11576722-11576722 on Build GRCh38
rs978324600 Chr.19: 11576608-11576608 on Build GRCh38
rs756448158 Chr.19: 11576732-11576733 on Build GRCh38
rs765343526 Chr.19: 11576720-11576720 on Build GRCh38
rs199580546 Chr.19: 11576719-11576719 on Build GRCh38
rs923204953 Chr.19: 11576697-11576697 on Build GRCh38
rs1007841729 Chr.19: 11576642-11576642 on Build GRCh38
rs750328133 Chr.19: 11576619-11576619 on Build GRCh38
rs776116121 Chr.19: 11576703-11576703 on Build GRCh38
rs182019242 Chr.19: 11576599-11576599 on Build GRCh38
rs201716955 Chr.19: 11576557-11576557 on Build GRCh38

View Full Product Details