Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81891314-81891559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALYREF
Entrez Gene ID
10189
Gene Name
Aly/REF export factor
Gene Aliases
ALY, ALY/REF, BEF, REF, THOC4
UniGene
Hs.534385
Gene Chromosome Location
Chr.17: 81887835-81891622 on Build GRCh38
Transcripts
NM_005782.3 NM_001289414.1

Gene Symbol
ANAPC11
Entrez Gene ID
51529
Gene Name
anaphase promoting complex subunit 11
Gene Aliases
APC11, Apc11p, HSPC214
UniGene
Hs.534456
Gene Chromosome Location
Chr.17: 81890790-81900533 on Build GRCh38
Transcripts
NM_005782.3 NM_001289414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381156 c.89G>A p.R30Q Chr.17: 81891492-81891492 on Build GRCh38
1666162 c.53G>A p.S18N Chr.17: 81891507-81891507 on Build GRCh38
6381157 c.68G>A p.R23Q Chr.17: 81891492-81891492 on Build GRCh38
5599066 c.45G>A p.L15L Chr.17: 81891515-81891515 on Build GRCh38
5729829 c.84C>T p.G28G Chr.17: 81891476-81891476 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1024058040 Chr.17: 81891441-81891441 on Build GRCh38
rs1035044135 Chr.17: 81891463-81891463 on Build GRCh38
rs758291181 Chr.17: 81891388-81891388 on Build GRCh38
rs889640750 Chr.17: 81891462-81891485 on Build GRCh38
rs893747241 Chr.17: 81891335-81891335 on Build GRCh38
rs752647095 Chr.17: 81891340-81891340 on Build GRCh38
rs951444306 Chr.17: 81891465-81891465 on Build GRCh38
rs963291405 Chr.17: 81891491-81891491 on Build GRCh38
rs567109460 Chr.17: 81891520-81891520 on Build GRCh38
rs969325896 Chr.17: 81891453-81891453 on Build GRCh38
rs983368408 Chr.17: 81891476-81891487 on Build GRCh38
rs777709104 Chr.17: 81891515-81891515 on Build GRCh38
rs975687462 Chr.17: 81891512-81891512 on Build GRCh38
rs758883369 Chr.17: 81891481-81891482 on Build GRCh38
rs555285564 Chr.17: 81891397-81891397 on Build GRCh38
rs867148285 Chr.17: 81891490-81891490 on Build GRCh38

View Full Product Details