Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91992832-91993093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP9
Entrez Gene ID
10142
Gene Name
A-kinase anchoring protein 9
Gene Aliases
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
UniGene
Hs.651221
Gene Chromosome Location
Chr.7: 91940867-92110673 on Build GRCh38
Transcripts
NM_005751.4 NM_147185.2 XM_017011642.1 XM_017011643.1 XM_017011644.1 XM_017011645.1 XM_017011646.1 XM_017011647.1 XM_017011648.1 XM_017011649.1 XM_017011650.1 XM_017011651.1 XM_017011652.1 XM_017011653.1 XM_017011654.1 XM_017011655.1 XM_017011656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7355078 c.510G>C p.E170D Chr.7: 91992989-91992989 on Build GRCh38
1698873 c.529G>A p.E177K Chr.7: 91993008-91993008 on Build GRCh38
1698871 c.480G>A p.M160I Chr.7: 91992959-91992959 on Build GRCh38
6189311 c.477G>T p.M159I Chr.7: 91992956-91992956 on Build GRCh38
1182399 c.407A>G p.E136G Chr.7: 91992886-91992886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56254439 Chr.7: 91992888-91992888 on Build GRCh38
rs371475691 Chr.7: 91992913-91992913 on Build GRCh38
rs749763833 Chr.7: 91993055-91993055 on Build GRCh38
rs150030669 Chr.7: 91992922-91992922 on Build GRCh38
rs779393285 Chr.7: 91993025-91993025 on Build GRCh38
rs201964732 Chr.7: 91992976-91992976 on Build GRCh38
rs746612786 Chr.7: 91992939-91992939 on Build GRCh38
rs367997608 Chr.7: 91992881-91992881 on Build GRCh38
rs769117540 Chr.7: 91993047-91993047 on Build GRCh38
rs774826824 Chr.7: 91993063-91993063 on Build GRCh38
rs373054959 Chr.7: 91993059-91993059 on Build GRCh38
rs768097461 Chr.7: 91992940-91992940 on Build GRCh38
rs745652672 Chr.7: 91992934-91992934 on Build GRCh38
rs199631609 Chr.7: 91993033-91993033 on Build GRCh38
rs376631476 Chr.7: 91993060-91993060 on Build GRCh38
rs778455688 Chr.7: 91993049-91993049 on Build GRCh38
rs759783541 Chr.7: 91992964-91992964 on Build GRCh38
rs758207667 Chr.7: 91992991-91992991 on Build GRCh38
rs761717651 Chr.7: 91992984-91992984 on Build GRCh38
rs773796684 Chr.7: 91992945-91992945 on Build GRCh38
rs564051054 Chr.7: 91992877-91992877 on Build GRCh38
rs148852801 Chr.7: 91992971-91992971 on Build GRCh38
rs755140795 Chr.7: 91992910-91992910 on Build GRCh38
rs779798051 Chr.7: 91992999-91992999 on Build GRCh38
rs185898118 Chr.7: 91992920-91992920 on Build GRCh38
rs142632541 Chr.7: 91992943-91992943 on Build GRCh38
rs766721210 Chr.7: 91992893-91992893 on Build GRCh38
rs375182237 Chr.7: 91992941-91992941 on Build GRCh38
rs780623014 Chr.7: 91993032-91993032 on Build GRCh38
rs896283872 Chr.7: 91993036-91993036 on Build GRCh38
rs753077637 Chr.7: 91993014-91993014 on Build GRCh38
rs753770347 Chr.7: 91992917-91992917 on Build GRCh38
rs527401771 Chr.7: 91992958-91992958 on Build GRCh38
rs138567837 Chr.7: 91992935-91992935 on Build GRCh38
rs759745243 Chr.7: 91993069-91993069 on Build GRCh38
rs774737080 Chr.7: 91992962-91992962 on Build GRCh38
rs74729008 Chr.7: 91993020-91993020 on Build GRCh38
rs376010972 Chr.7: 91992950-91992950 on Build GRCh38
rs751250535 Chr.7: 91993003-91993003 on Build GRCh38
rs779689614 Chr.7: 91992936-91992936 on Build GRCh38
rs751764031 Chr.7: 91992906-91992906 on Build GRCh38
rs200448722 Chr.7: 91992873-91992873 on Build GRCh38
rs750921440 Chr.7: 91993026-91993026 on Build GRCh38
rs539375675 Chr.7: 91992988-91992988 on Build GRCh38
rs144888041 Chr.7: 91992989-91992989 on Build GRCh38

View Full Product Details