Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122768903-122769150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENR
Entrez Gene ID
8562
Gene Name
density regulated re-initiation and release factor
Gene Aliases
DRP, DRP1, SMAP-3
UniGene
Hs.22393
Gene Chromosome Location
Chr.12: 122752821-122771064 on Build GRCh38
Transcripts
NM_003677.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6642535 c.571G>A p.E191K Chr.12: 122769052-122769052 on Build GRCh38
6642534 c.583G>A p.E195K Chr.12: 122769064-122769064 on Build GRCh38
415983 c.577C>A p.L193I Chr.12: 122769058-122769058 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374643112 Chr.12: 122768954-122768954 on Build GRCh38
rs201139163 Chr.12: 122768967-122768967 on Build GRCh38
rs746477224 Chr.12: 122768941-122768941 on Build GRCh38
rs773288960 Chr.12: 122769117-122769117 on Build GRCh38
rs373554836 Chr.12: 122769094-122769094 on Build GRCh38
rs769460847 Chr.12: 122768946-122768946 on Build GRCh38
rs770497545 Chr.12: 122768942-122768942 on Build GRCh38
rs768484434 Chr.12: 122769055-122769055 on Build GRCh38
rs1038937453 Chr.12: 122769090-122769093 on Build GRCh38
rs185706767 Chr.12: 122769051-122769051 on Build GRCh38
rs528184883 Chr.12: 122768945-122768945 on Build GRCh38
rs755425262 Chr.12: 122768998-122768998 on Build GRCh38
rs766587962 Chr.12: 122769125-122769125 on Build GRCh38
rs144554963 Chr.12: 122768949-122768949 on Build GRCh38
rs754193468 Chr.12: 122768991-122768991 on Build GRCh38
rs201636319 Chr.12: 122768943-122768943 on Build GRCh38
rs190514396 Chr.12: 122769090-122769090 on Build GRCh38
rs749029040 Chr.12: 122769052-122769052 on Build GRCh38
rs375401513 Chr.12: 122769115-122769115 on Build GRCh38
rs745382001 Chr.12: 122769030-122769030 on Build GRCh38
rs772034231 Chr.12: 122769116-122769116 on Build GRCh38
rs532771001 Chr.12: 122769000-122769000 on Build GRCh38
rs760679566 Chr.12: 122769119-122769119 on Build GRCh38
rs780811446 Chr.12: 122769013-122769013 on Build GRCh38
rs761018208 Chr.12: 122768969-122768969 on Build GRCh38
rs756692627 Chr.12: 122769002-122769002 on Build GRCh38
rs369866831 Chr.12: 122768956-122768956 on Build GRCh38
rs770468881 Chr.12: 122769107-122769111 on Build GRCh38
rs779570993 Chr.12: 122768930-122768930 on Build GRCh38
rs777329748 Chr.12: 122768955-122768955 on Build GRCh38
rs367903795 Chr.12: 122768929-122768929 on Build GRCh38
rs867781217 Chr.12: 122768978-122768978 on Build GRCh38
rs554942000 Chr.12: 122769101-122769101 on Build GRCh38
rs748690315 Chr.12: 122769011-122769012 on Build GRCh38
rs550965293 Chr.12: 122769045-122769045 on Build GRCh38

View Full Product Details