Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56372775-56373006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKFN1
Entrez Gene ID
162282
Gene Name
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases
-
UniGene
Hs.673040
Gene Chromosome Location
Chr.17: 56110935-56517016 on Build GRCh38
Transcripts
NM_153228.2 XM_006721728.3 XM_011524428.2 XM_011524429.2 XM_011524430.2 XM_011524431.2 XM_017024263.1 XM_017024264.1 XM_017024265.1 XM_017024266.1 XM_017024267.1 XM_017024268.1 XM_017024269.1 XM_017024270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1384581 c.742C>T p.L248L Chr.17: 56372777-56372777 on Build GRCh38
3890005 c.748G>A p.A250T Chr.17: 56372783-56372783 on Build GRCh38
4068001 c.760A>G p.R254G Chr.17: 56372795-56372795 on Build GRCh38
5508456 c.792C>A p.G264G Chr.17: 56372827-56372827 on Build GRCh38
6389636 c.805A>G p.R269G Chr.17: 56372840-56372840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763573020 Chr.17: 56372835-56372835 on Build GRCh38
rs997102112 Chr.17: 56372951-56372951 on Build GRCh38
rs958098165 Chr.17: 56372974-56372974 on Build GRCh38
rs766486507 Chr.17: 56372801-56372801 on Build GRCh38
rs1007861738 Chr.17: 56372898-56372898 on Build GRCh38
rs375674452 Chr.17: 56372884-56372884 on Build GRCh38
rs747961421 Chr.17: 56372883-56372883 on Build GRCh38
rs554885540 Chr.17: 56372952-56372952 on Build GRCh38
rs143082022 Chr.17: 56372813-56372813 on Build GRCh38
rs760059035 Chr.17: 56372803-56372803 on Build GRCh38
rs753297681 Chr.17: 56372910-56372910 on Build GRCh38
rs1008084667 Chr.17: 56372916-56372917 on Build GRCh38
rs778618702 Chr.17: 56372838-56372838 on Build GRCh38
rs1019553175 Chr.17: 56372902-56372902 on Build GRCh38
rs183259152 Chr.17: 56372935-56372935 on Build GRCh38
rs372385838 Chr.17: 56372873-56372873 on Build GRCh38
rs750154030 Chr.17: 56372849-56372849 on Build GRCh38
rs372503192 Chr.17: 56372885-56372885 on Build GRCh38
rs531017481 Chr.17: 56372826-56372826 on Build GRCh38
rs75135617 Chr.17: 56372859-56372859 on Build GRCh38
rs752022371 Chr.17: 56372802-56372802 on Build GRCh38
rs998528709 Chr.17: 56372885-56372898 on Build GRCh38
rs781056530 Chr.17: 56372882-56372882 on Build GRCh38
rs898376945 Chr.17: 56372854-56372854 on Build GRCh38
rs746883633 Chr.17: 56372872-56372872 on Build GRCh38
rs879682665 Chr.17: 56372917-56372917 on Build GRCh38
rs1045909617 Chr.17: 56372799-56372799 on Build GRCh38
rs1028953251 Chr.17: 56372965-56372965 on Build GRCh38
rs760218269 Chr.17: 56372832-56372832 on Build GRCh38

View Full Product Details