Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36151525-36151796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7A1
Entrez Gene ID
1346
Gene Name
cytochrome c oxidase subunit 7A1
Gene Aliases
COX7A, COX7AH, COX7AM
UniGene
Hs.421621
Gene Chromosome Location
Chr.19: 36150922-36152869 on Build GRCh38
Transcripts
NM_001864.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5855567 c.119C>T p.P40L Chr.19: 36151530-36151530 on Build GRCh38
3199575 c.105_107delGGA p.E35delE Chr.19: 36151542-36151544 on Build GRCh38
5855568 c.50C>T p.T17I Chr.19: 36151721-36151721 on Build GRCh38
6233534 c.102+10C>G p.? Chr.19: 36151659-36151659 on Build GRCh38
4281989 c.102+2T>G p.? Chr.19: 36151667-36151667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371763146 Chr.19: 36151653-36151653 on Build GRCh38
rs757007537 Chr.19: 36151596-36151619 on Build GRCh38
rs375173190 Chr.19: 36151650-36151650 on Build GRCh38
rs771394071 Chr.19: 36151697-36151697 on Build GRCh38
rs796346815 Chr.19: 36151659-36151660 on Build GRCh38
rs17882022 Chr.19: 36151658-36151658 on Build GRCh38
rs769604961 Chr.19: 36151557-36151557 on Build GRCh38
rs3214131 Chr.19: 36151666-36151667 on Build GRCh38
rs748684336 Chr.19: 36151656-36151657 on Build GRCh38
rs1039897846 Chr.19: 36151690-36151690 on Build GRCh38
rs759703484 Chr.19: 36151618-36151618 on Build GRCh38
rs751451644 Chr.19: 36151628-36151628 on Build GRCh38
rs774812314 Chr.19: 36151559-36151559 on Build GRCh38
rs958591246 Chr.19: 36151549-36151549 on Build GRCh38
rs779840985 Chr.19: 36151668-36151668 on Build GRCh38
rs766426302 Chr.19: 36151591-36151591 on Build GRCh38
rs201977124 Chr.19: 36151767-36151767 on Build GRCh38
rs1042123290 Chr.19: 36151759-36151759 on Build GRCh38
rs765489898 Chr.19: 36151619-36151619 on Build GRCh38
rs761753337 Chr.19: 36151660-36151661 on Build GRCh38
rs530285319 Chr.19: 36151678-36151680 on Build GRCh38
rs754976929 Chr.19: 36151663-36151664 on Build GRCh38
rs745934643 Chr.19: 36151719-36151719 on Build GRCh38
rs112834485 Chr.19: 36151659-36151660 on Build GRCh38
rs753472990 Chr.19: 36151669-36151669 on Build GRCh38
rs776697632 Chr.19: 36151594-36151594 on Build GRCh38
rs946642034 Chr.19: 36151771-36151771 on Build GRCh38
rs376001720 Chr.19: 36151666-36151666 on Build GRCh38
rs980569290 Chr.19: 36151659-36151659 on Build GRCh38
rs570117682 Chr.19: 36151606-36151606 on Build GRCh38
rs767562238 Chr.19: 36151765-36151765 on Build GRCh38
rs781713923 Chr.19: 36151714-36151714 on Build GRCh38
rs780576270 Chr.19: 36151548-36151548 on Build GRCh38
rs371700119 Chr.19: 36151656-36151656 on Build GRCh38
rs763921400 Chr.19: 36151623-36151623 on Build GRCh38
rs750588406 Chr.19: 36151650-36151651 on Build GRCh38
rs755756129 Chr.19: 36151667-36151667 on Build GRCh38
rs758278756 Chr.19: 36151622-36151622 on Build GRCh38
rs971014309 Chr.19: 36151634-36151634 on Build GRCh38
rs747380164 Chr.19: 36151691-36151691 on Build GRCh38
rs750444414 Chr.19: 36151766-36151766 on Build GRCh38
rs767322790 Chr.19: 36151651-36151651 on Build GRCh38
rs199557841 Chr.19: 36151653-36151660 on Build GRCh38
rs372662024 Chr.19: 36151566-36151566 on Build GRCh38
rs751337144 Chr.19: 36151554-36151555 on Build GRCh38
rs763192376 Chr.19: 36151744-36151744 on Build GRCh38
rs768780058 Chr.19: 36151752-36151752 on Build GRCh38
rs745397036 Chr.19: 36151651-36151651 on Build GRCh38
rs933983257 Chr.19: 36151703-36151703 on Build GRCh38
rs773464168 Chr.19: 36151654-36151654 on Build GRCh38
rs867102142 Chr.19: 36151715-36151715 on Build GRCh38
rs779060338 Chr.19: 36151664-36151665 on Build GRCh38
rs745444096 Chr.19: 36151553-36151553 on Build GRCh38
rs2285598 Chr.19: 36151652-36151652 on Build GRCh38
rs753926083 Chr.19: 36151661-36151662 on Build GRCh38
rs749388258 Chr.19: 36151654-36151660 on Build GRCh38
rs755492987 Chr.19: 36151655-36151656 on Build GRCh38
rs34667074 Chr.19: 36151559-36151560 on Build GRCh38
rs753185829 Chr.19: 36151665-36151666 on Build GRCh38
rs761764097 Chr.19: 36151659-36151659 on Build GRCh38
rs751339646 Chr.19: 36151657-36151657 on Build GRCh38
rs149425283 Chr.19: 36151735-36151735 on Build GRCh38
rs769803673 Chr.19: 36151729-36151729 on Build GRCh38
rs757144028 Chr.19: 36151635-36151635 on Build GRCh38
rs752457786 Chr.19: 36151621-36151621 on Build GRCh38
rs754593590 Chr.19: 36151670-36151670 on Build GRCh38
rs761557986 Chr.19: 36151760-36151760 on Build GRCh38
rs555443322 Chr.19: 36151689-36151689 on Build GRCh38
rs745507376 Chr.19: 36151658-36151659 on Build GRCh38
rs776668775 Chr.19: 36151655-36151655 on Build GRCh38
rs780812653 Chr.19: 36151641-36151641 on Build GRCh38
rs763986935 Chr.19: 36151549-36151549 on Build GRCh38
rs2285599 Chr.19: 36151660-36151660 on Build GRCh38
rs760657515 Chr.19: 36151578-36151578 on Build GRCh38
rs774176689 Chr.19: 36151756-36151756 on Build GRCh38

View Full Product Details