Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154584977-154585232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAR
Entrez Gene ID
103
Gene Name
adenosine deaminase, RNA specific
Gene Aliases
ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
UniGene
Hs.12341
Gene Chromosome Location
Chr.1: 154582057-154628016 on Build GRCh38
Transcripts
NM_001025107.2 NM_001111.4 NM_001193495.1 NM_015840.3 NM_015841.3 XM_006711109.2 XM_006711111.3 XM_006711112.2 XM_006711113.2 XM_011509060.1 XM_011509061.2 XM_011509062.1 XM_017000037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022834 c.3448C>T p.R1150W Chr.1: 154585039-154585039 on Build GRCh38
3802036 c.3477G>T p.K1159N Chr.1: 154585010-154585010 on Build GRCh38
6567142 c.3616C>T p.L1206F Chr.1: 154585000-154585000 on Build GRCh38
4400376 c.3462C>T p.S1154S Chr.1: 154585025-154585025 on Build GRCh38
3802037 c.3606G>T p.K1202N Chr.1: 154585010-154585010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561276347 Chr.1: 154585148-154585148 on Build GRCh38
rs781290471 Chr.1: 154585161-154585161 on Build GRCh38
rs531922411 Chr.1: 154585207-154585207 on Build GRCh38
rs542283847 Chr.1: 154585111-154585111 on Build GRCh38
rs376661636 Chr.1: 154585167-154585167 on Build GRCh38
rs556424333 Chr.1: 154585077-154585077 on Build GRCh38
rs758253421 Chr.1: 154585025-154585025 on Build GRCh38
rs747744936 Chr.1: 154585087-154585087 on Build GRCh38
rs201327808 Chr.1: 154585164-154585164 on Build GRCh38
rs759837531 Chr.1: 154585175-154585175 on Build GRCh38
rs557095887 Chr.1: 154585126-154585126 on Build GRCh38
rs756399730 Chr.1: 154585193-154585193 on Build GRCh38
rs779950072 Chr.1: 154585038-154585038 on Build GRCh38
rs1044845711 Chr.1: 154585024-154585024 on Build GRCh38
rs1006116725 Chr.1: 154585033-154585033 on Build GRCh38
rs775046967 Chr.1: 154585090-154585092 on Build GRCh38
rs746750625 Chr.1: 154585056-154585056 on Build GRCh38
rs752973159 Chr.1: 154585187-154585187 on Build GRCh38
rs768458733 Chr.1: 154585060-154585060 on Build GRCh38
rs970217979 Chr.1: 154585152-154585152 on Build GRCh38
rs771736098 Chr.1: 154585089-154585100 on Build GRCh38
rs368716872 Chr.1: 154585171-154585171 on Build GRCh38
rs574813263 Chr.1: 154585086-154585086 on Build GRCh38
rs372228353 Chr.1: 154585088-154585088 on Build GRCh38
rs9427094 Chr.1: 154585209-154585209 on Build GRCh38
rs745866663 Chr.1: 154585020-154585020 on Build GRCh38
rs999172070 Chr.1: 154585039-154585039 on Build GRCh38
rs887707722 Chr.1: 154585027-154585027 on Build GRCh38
rs766763044 Chr.1: 154585168-154585168 on Build GRCh38
rs199583213 Chr.1: 154585183-154585183 on Build GRCh38
rs764325867 Chr.1: 154585196-154585196 on Build GRCh38
rs997889284 Chr.1: 154585094-154585094 on Build GRCh38
rs560401349 Chr.1: 154585125-154585125 on Build GRCh38
rs1030430888 Chr.1: 154585121-154585121 on Build GRCh38
rs760260332 Chr.1: 154585184-154585185 on Build GRCh38
rs200649075 Chr.1: 154585198-154585198 on Build GRCh38

View Full Product Details