Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45848617-45848854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT8
Entrez Gene ID
10005
Gene Name
acyl-CoA thioesterase 8
Gene Aliases
HNAACTE, NAP1, PTE-1, PTE-2, PTE1, PTE2, hACTE-III, hTE
UniGene
Hs.444776
Gene Chromosome Location
Chr.20: 45841721-45857409 on Build GRCh38
Transcripts
NM_005469.3 XM_005260239.3 XM_011528480.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904036 c.263-8C>T p.? Chr.20: 45848683-45848683 on Build GRCh38
3547099 c.305G>A p.R102Q Chr.20: 45848633-45848633 on Build GRCh38
298315 c.273_274insTGATT p.L92fs*1 Chr.20: 45848664-45848665 on Build GRCh38
6592337 c.315G>A p.S105S Chr.20: 45848623-45848623 on Build GRCh38
5057356 c.277C>T p.P93S Chr.20: 45848661-45848661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375697034 Chr.20: 45848690-45848690 on Build GRCh38
rs371617984 Chr.20: 45848639-45848639 on Build GRCh38
rs1019365876 Chr.20: 45848818-45848818 on Build GRCh38
rs553157848 Chr.20: 45848728-45848728 on Build GRCh38
rs34282076 Chr.20: 45848692-45848693 on Build GRCh38
rs749089125 Chr.20: 45848719-45848719 on Build GRCh38
rs900304740 Chr.20: 45848820-45848820 on Build GRCh38
rs995109600 Chr.20: 45848794-45848794 on Build GRCh38
rs897162924 Chr.20: 45848714-45848714 on Build GRCh38
rs772020067 Chr.20: 45848640-45848640 on Build GRCh38
rs1038553493 Chr.20: 45848712-45848712 on Build GRCh38
rs375537676 Chr.20: 45848700-45848700 on Build GRCh38
rs138226225 Chr.20: 45848669-45848669 on Build GRCh38
rs751125055 Chr.20: 45848681-45848681 on Build GRCh38
rs771477337 Chr.20: 45848822-45848822 on Build GRCh38
rs138917356 Chr.20: 45848814-45848814 on Build GRCh38
rs766159231 Chr.20: 45848680-45848680 on Build GRCh38
rs941649323 Chr.20: 45848707-45848707 on Build GRCh38
rs899426547 Chr.20: 45848787-45848787 on Build GRCh38
rs1051198935 Chr.20: 45848773-45848773 on Build GRCh38
rs938195368 Chr.20: 45848755-45848755 on Build GRCh38
rs777633776 Chr.20: 45848715-45848715 on Build GRCh38
rs370051307 Chr.20: 45848724-45848724 on Build GRCh38
rs761078019 Chr.20: 45848663-45848663 on Build GRCh38
rs755995482 Chr.20: 45848702-45848702 on Build GRCh38
rs929941735 Chr.20: 45848752-45848752 on Build GRCh38
rs975699628 Chr.20: 45848653-45848653 on Build GRCh38
rs201318577 Chr.20: 45848670-45848670 on Build GRCh38
rs200128490 Chr.20: 45848682-45848682 on Build GRCh38
rs764609968 Chr.20: 45848668-45848668 on Build GRCh38
rs772746705 Chr.20: 45848696-45848696 on Build GRCh38
rs1046593450 Chr.20: 45848738-45848738 on Build GRCh38
rs796721209 Chr.20: 45848744-45848744 on Build GRCh38
rs776099929 Chr.20: 45848647-45848647 on Build GRCh38
rs113422635 Chr.20: 45848797-45848797 on Build GRCh38
rs767665213 Chr.20: 45848716-45848718 on Build GRCh38
rs905398131 Chr.20: 45848749-45848749 on Build GRCh38
rs150217048 Chr.20: 45848798-45848798 on Build GRCh38

View Full Product Details