Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167809483-167809754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY10
Entrez Gene ID
55811
Gene Name
adenylate cyclase 10, soluble
Gene Aliases
HCA2, HEL-S-7a, SAC, SACI, Sacy, hsAC
UniGene
Hs.320892
Gene Chromosome Location
Chr.1: 167809080-167915556 on Build GRCh38
Transcripts
NM_001167749.2 NM_001297772.1 NM_018417.5 XM_006711449.3 XM_011509760.2 XM_011509762.2 XM_011509763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
278659 c.4815C>T p.T1605T Chr.1: 167809696-167809696 on Build GRCh38
1336009 c.4790_4791insA p.N1597fs*8 Chr.1: 167809720-167809721 on Build GRCh38
5480925 c.4812T>A p.N1604K Chr.1: 167809699-167809699 on Build GRCh38
1472935 c.4826A>C p.H1609P Chr.1: 167809685-167809685 on Build GRCh38
3789052 c.4774C>G p.Q1592E Chr.1: 167809737-167809737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369689948 Chr.1: 167809627-167809627 on Build GRCh38
rs112911097 Chr.1: 167809689-167809689 on Build GRCh38
rs1009364935 Chr.1: 167809526-167809526 on Build GRCh38
rs555993450 Chr.1: 167809611-167809611 on Build GRCh38
rs753528374 Chr.1: 167809709-167809709 on Build GRCh38
rs548840962 Chr.1: 167809583-167809583 on Build GRCh38
rs374079289 Chr.1: 167809645-167809645 on Build GRCh38
rs377762633 Chr.1: 167809661-167809661 on Build GRCh38
rs1045726033 Chr.1: 167809618-167809618 on Build GRCh38
rs755107850 Chr.1: 167809550-167809550 on Build GRCh38
rs937408774 Chr.1: 167809629-167809629 on Build GRCh38
rs753500756 Chr.1: 167809720-167809721 on Build GRCh38
rs774444517 Chr.1: 167809675-167809675 on Build GRCh38
rs911038551 Chr.1: 167809576-167809576 on Build GRCh38
rs767453932 Chr.1: 167809693-167809693 on Build GRCh38
rs955097828 Chr.1: 167809529-167809529 on Build GRCh38
rs75466950 Chr.1: 167809626-167809626 on Build GRCh38
rs773450284 Chr.1: 167809643-167809643 on Build GRCh38
rs527356749 Chr.1: 167809527-167809527 on Build GRCh38
rs907337160 Chr.1: 167809623-167809623 on Build GRCh38
rs764869983 Chr.1: 167809599-167809599 on Build GRCh38
rs3213588 Chr.1: 167809606-167809606 on Build GRCh38
rs146952021 Chr.1: 167809696-167809696 on Build GRCh38
rs769957178 Chr.1: 167809631-167809631 on Build GRCh38
rs1038712521 Chr.1: 167809608-167809608 on Build GRCh38
rs985328086 Chr.1: 167809541-167809541 on Build GRCh38
rs1055918254 Chr.1: 167809669-167809669 on Build GRCh38
rs1035063055 Chr.1: 167809676-167809676 on Build GRCh38
rs373395570 Chr.1: 167809705-167809705 on Build GRCh38
rs548412121 Chr.1: 167809610-167809610 on Build GRCh38
rs890465213 Chr.1: 167809522-167809522 on Build GRCh38
rs879191093 Chr.1: 167809592-167809592 on Build GRCh38
rs913636305 Chr.1: 167809721-167809721 on Build GRCh38
rs201892899 Chr.1: 167809695-167809695 on Build GRCh38

View Full Product Details