Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30066867-30067094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOA
Entrez Gene ID
226
Gene Name
aldolase, fructose-bisphosphate A
Gene Aliases
ALDA, GSD12, HEL-S-87p
UniGene
Hs.513490
Gene Chromosome Location
Chr.16: 30053090-30070420 on Build GRCh38
Transcripts
NM_000034.3 NM_001127617.2 NM_001243177.1 NM_184041.2 NM_184043.2 XM_011545768.2

SNP Detail

SNP ID Chromosome Location
rs920402964 Chr.16: 30067044-30067044 on Build GRCh38
rs754143271 Chr.16: 30066998-30066998 on Build GRCh38
rs764462408 Chr.16: 30066996-30066996 on Build GRCh38
rs971402340 Chr.16: 30066939-30066939 on Build GRCh38
rs757710745 Chr.16: 30066906-30066906 on Build GRCh38
rs199775687 Chr.16: 30067011-30067011 on Build GRCh38
rs903764420 Chr.16: 30067060-30067060 on Build GRCh38
rs768714573 Chr.16: 30067062-30067062 on Build GRCh38
rs552061511 Chr.16: 30066907-30066907 on Build GRCh38
rs1036530879 Chr.16: 30066955-30066956 on Build GRCh38
rs768781534 Chr.16: 30066970-30066970 on Build GRCh38
rs146835556 Chr.16: 30066908-30066908 on Build GRCh38
rs760860128 Chr.16: 30066977-30066977 on Build GRCh38
rs780378047 Chr.16: 30066927-30066927 on Build GRCh38
rs531033350 Chr.16: 30066919-30066919 on Build GRCh38
rs750912887 Chr.16: 30067008-30067008 on Build GRCh38
rs765795855 Chr.16: 30066983-30066983 on Build GRCh38
rs768538661 Chr.16: 30066942-30066942 on Build GRCh38
rs769800173 Chr.16: 30066961-30066961 on Build GRCh38
rs754372047 Chr.16: 30066899-30066899 on Build GRCh38
rs914136423 Chr.16: 30066938-30066938 on Build GRCh38
rs535217608 Chr.16: 30067053-30067053 on Build GRCh38
rs546976628 Chr.16: 30067072-30067072 on Build GRCh38
rs772036398 Chr.16: 30066924-30066924 on Build GRCh38
rs776529531 Chr.16: 30066943-30066943 on Build GRCh38
rs530878836 Chr.16: 30067073-30067073 on Build GRCh38
rs567803172 Chr.16: 30067000-30067000 on Build GRCh38
rs759070394 Chr.16: 30066994-30066994 on Build GRCh38
rs762292824 Chr.16: 30067026-30067027 on Build GRCh38
rs779460013 Chr.16: 30067004-30067004 on Build GRCh38
rs779963072 Chr.16: 30067047-30067047 on Build GRCh38
rs973485583 Chr.16: 30066965-30066965 on Build GRCh38
rs772853499 Chr.16: 30066971-30066971 on Build GRCh38
rs751275868 Chr.16: 30066987-30066987 on Build GRCh38
rs559971583 Chr.16: 30066901-30066901 on Build GRCh38
rs549434187 Chr.16: 30066958-30066958 on Build GRCh38
rs747173726 Chr.16: 30066934-30066934 on Build GRCh38
rs3112553 Chr.16: 30067043-30067043 on Build GRCh38
rs926663650 Chr.16: 30066944-30066944 on Build GRCh38
rs758552797 Chr.16: 30067032-30067032 on Build GRCh38
rs762591392 Chr.16: 30066972-30066972 on Build GRCh38

View Full Product Details