Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45397834-45398091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP27
Entrez Gene ID
201176
Gene Name
Rho GTPase activating protein 27
Gene Aliases
CAMGAP1, PP905, SH3D20, SH3P20
UniGene
Hs.205326
Gene Chromosome Location
Chr.17: 45393902-45432916 on Build GRCh38
Transcripts
NM_001282290.1 NM_199282.2 XM_005257128.3 XM_006721745.2 XM_006721747.2 XM_006721750.3 XM_011524468.1 XM_011524469.1 XM_011524470.2 XM_011524471.1 XM_011524472.2 XM_011524474.1 XM_017024311.1 XM_017024312.1 XM_017024313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413888 c.1810C>T p.H604Y Chr.17: 45397981-45397981 on Build GRCh38
6413887 c.787C>T p.H263Y Chr.17: 45397981-45397981 on Build GRCh38
6413634 c.1816G>T p.A606S Chr.17: 45397975-45397975 on Build GRCh38
6413633 c.793G>T p.A265S Chr.17: 45397975-45397975 on Build GRCh38
980147 c.761A>G p.D254G Chr.17: 45398007-45398007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745407695 Chr.17: 45398015-45398015 on Build GRCh38
rs35717383 Chr.17: 45398035-45398035 on Build GRCh38
rs780665476 Chr.17: 45397898-45397898 on Build GRCh38
rs957118898 Chr.17: 45397889-45397890 on Build GRCh38
rs11653604 Chr.17: 45397878-45397878 on Build GRCh38
rs758454805 Chr.17: 45397902-45397902 on Build GRCh38
rs542465220 Chr.17: 45397894-45397894 on Build GRCh38
rs777691376 Chr.17: 45397915-45397915 on Build GRCh38
rs759447924 Chr.17: 45397967-45397967 on Build GRCh38
rs138416055 Chr.17: 45397954-45397954 on Build GRCh38
rs200137454 Chr.17: 45397924-45397924 on Build GRCh38
rs907339954 Chr.17: 45397860-45397860 on Build GRCh38
rs760608101 Chr.17: 45397957-45397957 on Build GRCh38
rs367987670 Chr.17: 45397976-45397976 on Build GRCh38
rs368752990 Chr.17: 45398044-45398044 on Build GRCh38
rs139052373 Chr.17: 45398037-45398037 on Build GRCh38
rs757388391 Chr.17: 45398065-45398066 on Build GRCh38
rs763585961 Chr.17: 45398004-45398004 on Build GRCh38
rs142144632 Chr.17: 45397907-45397907 on Build GRCh38
rs1007694625 Chr.17: 45397987-45397987 on Build GRCh38
rs1032652194 Chr.17: 45397876-45397876 on Build GRCh38
rs774822507 Chr.17: 45397908-45397908 on Build GRCh38
rs777538881 Chr.17: 45397882-45397882 on Build GRCh38
rs776504840 Chr.17: 45397966-45397966 on Build GRCh38
rs201061334 Chr.17: 45397926-45397926 on Build GRCh38
rs909182005 Chr.17: 45398040-45398040 on Build GRCh38
rs776662839 Chr.17: 45398061-45398061 on Build GRCh38
rs76353181 Chr.17: 45397964-45397964 on Build GRCh38
rs150663614 Chr.17: 45398008-45398008 on Build GRCh38
rs756795398 Chr.17: 45397895-45397895 on Build GRCh38
rs141908251 Chr.17: 45398003-45398003 on Build GRCh38
rs138914274 Chr.17: 45398009-45398009 on Build GRCh38
rs868396784 Chr.17: 45398054-45398054 on Build GRCh38
rs751846414 Chr.17: 45397916-45397916 on Build GRCh38
rs765199268 Chr.17: 45397970-45397970 on Build GRCh38
rs757413172 Chr.17: 45397923-45397923 on Build GRCh38
rs779879418 Chr.17: 45397952-45397952 on Build GRCh38
rs779617774 Chr.17: 45398030-45398030 on Build GRCh38
rs200660366 Chr.17: 45397965-45397965 on Build GRCh38
rs149430480 Chr.17: 45398024-45398024 on Build GRCh38
rs748017487 Chr.17: 45398068-45398068 on Build GRCh38

View Full Product Details