Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20455541-20455806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APEX1
Entrez Gene ID
328
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
Gene Aliases
APE, APE1, APEN, APEX, APX, HAP1, REF1
UniGene
Hs.73722
Gene Chromosome Location
Chr.14: 20455131-20457772 on Build GRCh38
Transcripts
NM_001244249.1 NM_001641.3 NM_080648.2 NM_080649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049724 c.9G>A p.K3K Chr.14: 20455654-20455654 on Build GRCh38
4049723 c.8A>G p.K3R Chr.14: 20455653-20455653 on Build GRCh38
432806 c.14G>C p.G5A Chr.14: 20455659-20455659 on Build GRCh38
2029124 c.20A>G p.K7R Chr.14: 20455665-20455665 on Build GRCh38
3494701 c.46G>A p.E16K Chr.14: 20455691-20455691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760532371 Chr.14: 20455731-20455731 on Build GRCh38
rs901653534 Chr.14: 20455724-20455724 on Build GRCh38
rs367585418 Chr.14: 20455695-20455695 on Build GRCh38
rs200470580 Chr.14: 20455625-20455625 on Build GRCh38
rs772548837 Chr.14: 20455776-20455776 on Build GRCh38
rs778607554 Chr.14: 20455692-20455692 on Build GRCh38
rs775333541 Chr.14: 20455614-20455614 on Build GRCh38
rs544923016 Chr.14: 20455743-20455744 on Build GRCh38
rs750720621 Chr.14: 20455750-20455750 on Build GRCh38
rs750669164 Chr.14: 20455639-20455639 on Build GRCh38
rs781707896 Chr.14: 20455763-20455763 on Build GRCh38
rs759108578 Chr.14: 20455606-20455606 on Build GRCh38
rs752031447 Chr.14: 20455650-20455650 on Build GRCh38
rs758446218 Chr.14: 20455642-20455642 on Build GRCh38
rs561258433 Chr.14: 20455729-20455729 on Build GRCh38
rs755473021 Chr.14: 20455663-20455663 on Build GRCh38
rs753201737 Chr.14: 20455770-20455770 on Build GRCh38
rs745532557 Chr.14: 20455774-20455774 on Build GRCh38
rs765092491 Chr.14: 20455747-20455747 on Build GRCh38
rs375926125 Chr.14: 20455638-20455638 on Build GRCh38
rs780157628 Chr.14: 20455643-20455643 on Build GRCh38
rs771611421 Chr.14: 20455613-20455613 on Build GRCh38
rs748787250 Chr.14: 20455612-20455613 on Build GRCh38
rs761765372 Chr.14: 20455746-20455746 on Build GRCh38
rs375789982 Chr.14: 20455759-20455759 on Build GRCh38
rs542006752 Chr.14: 20455595-20455595 on Build GRCh38
rs201573557 Chr.14: 20455726-20455726 on Build GRCh38
rs763184333 Chr.14: 20455752-20455752 on Build GRCh38
rs972172172 Chr.14: 20455774-20455777 on Build GRCh38
rs776692597 Chr.14: 20455744-20455744 on Build GRCh38
rs748497625 Chr.14: 20455702-20455702 on Build GRCh38
rs770525472 Chr.14: 20455724-20455726 on Build GRCh38
rs1006213174 Chr.14: 20455785-20455785 on Build GRCh38
rs1054565866 Chr.14: 20455628-20455628 on Build GRCh38
rs770134762 Chr.14: 20455688-20455688 on Build GRCh38
rs760641928 Chr.14: 20455617-20455617 on Build GRCh38
rs1011661042 Chr.14: 20455583-20455583 on Build GRCh38
rs753618515 Chr.14: 20455627-20455627 on Build GRCh38
rs759994592 Chr.14: 20455768-20455769 on Build GRCh38
rs771685188 Chr.14: 20455784-20455784 on Build GRCh38
rs766402740 Chr.14: 20455758-20455758 on Build GRCh38
rs762136931 Chr.14: 20455633-20455633 on Build GRCh38
rs755453426 Chr.14: 20455761-20455761 on Build GRCh38
rs748454523 Chr.14: 20455672-20455672 on Build GRCh38
rs1050449911 Chr.14: 20455728-20455728 on Build GRCh38
rs778090711 Chr.14: 20455772-20455772 on Build GRCh38
rs202001645 Chr.14: 20455667-20455667 on Build GRCh38
rs2307485 Chr.14: 20455572-20455572 on Build GRCh38
rs1027965485 Chr.14: 20455601-20455601 on Build GRCh38

View Full Product Details