Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46333437-46333662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKZ
Entrez Gene ID
8525
Gene Name
diacylglycerol kinase zeta
Gene Aliases
DAGK5, DAGK6, DGK-ZETA, hDGKzeta
UniGene
Hs.502461
Gene Chromosome Location
Chr.11: 46332905-46380554 on Build GRCh38
Transcripts
NM_201532.2 XM_005253181.3

SNP Detail

SNP ID Chromosome Location
rs376682693 Chr.11: 46333620-46333620 on Build GRCh38
rs765048587 Chr.11: 46333474-46333474 on Build GRCh38
rs111324926 Chr.11: 46333522-46333522 on Build GRCh38
rs986375677 Chr.11: 46333641-46333641 on Build GRCh38
rs973150452 Chr.11: 46333602-46333602 on Build GRCh38
rs1035246826 Chr.11: 46333531-46333531 on Build GRCh38
rs920426550 Chr.11: 46333611-46333611 on Build GRCh38
rs762937344 Chr.11: 46333499-46333499 on Build GRCh38
rs1015014880 Chr.11: 46333598-46333598 on Build GRCh38
rs761728572 Chr.11: 46333457-46333457 on Build GRCh38
rs756129517 Chr.11: 46333527-46333527 on Build GRCh38
rs56044090 Chr.11: 46333465-46333465 on Build GRCh38
rs902146471 Chr.11: 46333462-46333462 on Build GRCh38
rs1025732059 Chr.11: 46333642-46333642 on Build GRCh38
rs953245478 Chr.11: 46333618-46333619 on Build GRCh38
rs995960300 Chr.11: 46333631-46333631 on Build GRCh38
rs962605733 Chr.11: 46333612-46333612 on Build GRCh38
rs1043901155 Chr.11: 46333489-46333489 on Build GRCh38
rs1006834830 Chr.11: 46333559-46333559 on Build GRCh38
rs1013897102 Chr.11: 46333569-46333569 on Build GRCh38
rs767563675 Chr.11: 46333514-46333514 on Build GRCh38
rs1018520384 Chr.11: 46333587-46333587 on Build GRCh38
rs117762930 Chr.11: 46333558-46333558 on Build GRCh38
rs892525780 Chr.11: 46333459-46333459 on Build GRCh38
rs773075223 Chr.11: 46333476-46333476 on Build GRCh38
rs369730059 Chr.11: 46333480-46333480 on Build GRCh38
rs961754483 Chr.11: 46333599-46333599 on Build GRCh38

View Full Product Details